Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574499_m1
          See other TDO2 GE Assays ›
          Gene Symbol:
          Tdo2
          Gene Name:
          tryptophan 2,3-dioxygenase
          Gene Aliases:
          TO, Tdo
          Chromosome Location:
          Chr.2: 180897059 - 180914919 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574499_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tdo2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022403.2 NP_071798.1 3 - 4 278 113
          GenBank mRNA BC089802.1 - 3 - 4 274 7370033 113
          M55167.1 - 3 - 4 278 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003928.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tdo2

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          tryptophan metabolic process
          tryptophan catabolic process to kynurenine
          tryptophan catabolic process to acetyl-CoA
          oxidation-reduction process
          tryptophan 2,3-dioxygenase activity
          amino acid binding
          oxygen binding
          heme binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P800916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872079 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161586 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P800916 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P800916 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P800916 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P800916 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700303201 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57814 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31323 - -
          Agilent Rat cDNA Microarray (G4105A) 700303201 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823898 - -
          Affymetrix Rat Gene 1.0 ST Array 10823900 - -
          Affymetrix Rat Gene 1.0 ST Array 10823896 - -
          Affymetrix Rat Gene 1.0 ST Array 10823893 - -
          Affymetrix Rat Gene 1.0 ST Array 10823901 - -
          Affymetrix Rat Gene 1.0 ST Array 10823894 - -
          Affymetrix Rat Gene 1.0 ST Array 10823899 - -
          Affymetrix Rat Gene 1.0 ST Array 10823895 - -
          Affymetrix Rat Gene 1.0 ST Array 10823892 - -
          Affymetrix Rat Gene 1.0 ST Array 10823891 - -
          Affymetrix Rat Gene 1.0 ST Array 10823902 - -
          Affymetrix Rat Gene 1.0 ST Array 10823897 - -
          Affymetrix Rat Gene 1.1 ST Array 10823890 - -
          Rat Genome 230 2.0 Array 1368720_at - -
          Rat Genome 230 2.0 Array 1368720_PM_at - -
          Affymetrix HT Rat Focus Array 1368720_PM_at - -
          Affymetrix HT Rat Focus Array 1368720_at - -
          Affymetrix Rat Clariom S Assay PSR0200026363.rn.1 TC0200003928.rn.1 RTA TC
          Rat Expression Set 230 1368720_at - -
          Rat Expression Set 230 1368720_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10823890 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21905017 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369462 - -
          RatRef-12 Expression BeadChip ILMN_61834 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline