Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574526_m1
          See other ACVRL1 GE Assays ›
          Gene Symbol:
          Acvrl1
          Gene Name:
          activin A receptor type II-like 1
          Gene Aliases:
          R-3, R3, SETHKIR
          Chromosome Location:
          Chr.7: 142770486 - 142787336 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574526_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acvrl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022441.2 NP_071886.1 9 - 10 1522 122
          XM_006242303.2 XP_006242365.1 10 - 11 1923 122
          XM_006242304.2 XP_006242366.1 10 - 11 2527 122
          XM_006242305.2 XP_006242367.1 9 - 10 2542 122
          XM_006242307.2 XP_006242369.1 9 - 10 2412 122
          XM_008765673.1 XP_008763895.1 10 - 11 2586 122
          GenBank mRNA BC083173.1 - 9 - 10 1522 7098096 122
          FQ223580.1 - 9 - 10 1564 122
          L36088.1 - 10 - 11 1937 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001864.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acvrl1

          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          in utero embryonic development
          negative regulation of endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          lymphangiogenesis
          blood vessel remodeling
          regulation of transcription, DNA-templated
          protein phosphorylation
          negative regulation of cell adhesion
          signal transduction
          transforming growth factor beta receptor signaling pathway
          blood circulation
          regulation of blood pressure
          negative regulation of cell proliferation
          negative regulation of endothelial cell migration
          positive regulation of pathway-restricted SMAD protein phosphorylation
          signal transduction by protein phosphorylation
          negative regulation of cell growth
          negative regulation of cell migration
          BMP signaling pathway
          positive regulation of BMP signaling pathway
          activin receptor signaling pathway
          wound healing, spreading of epidermal cells
          negative regulation of blood vessel endothelial cell migration
          negative regulation of endothelial cell differentiation
          positive regulation of endothelial cell differentiation
          positive regulation of angiogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          blood vessel morphogenesis
          protein heterooligomerization
          negative regulation of focal adhesion assembly
          lymphatic endothelial cell differentiation
          artery development
          venous blood vessel development
          endothelial tube morphogenesis
          retina vasculature development in camera-type eye
          cellular response to transforming growth factor beta stimulus
          cellular response to BMP stimulus
          negative regulation of DNA biosynthetic process
          protein serine/threonine kinase activity
          transmembrane receptor protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          transforming growth factor beta-activated receptor activity
          transforming growth factor beta receptor activity, type I
          ATP binding
          activin receptor activity, type I
          protein kinase binding
          SMAD binding
          metal ion binding
          activin binding
          transforming growth factor beta binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P771346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120248 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P771346 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P771346 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P771346 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P771346 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700282925 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702144068 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3903 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57662 - -
          Agilent Rat cDNA Microarray (G4105A) 702144068 - -
          Agilent Rat cDNA Microarray (G4105A) 700282925 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10899356 - -
          Affymetrix Rat Gene 1.0 ST Array 10899359 - -
          Affymetrix Rat Gene 1.0 ST Array 10899365 - -
          Affymetrix Rat Gene 1.0 ST Array 10899357 - -
          Affymetrix Rat Gene 1.0 ST Array 10899355 - -
          Affymetrix Rat Gene 1.0 ST Array 10899362 - -
          Affymetrix Rat Gene 1.0 ST Array 10899363 - -
          Affymetrix Rat Gene 1.0 ST Array 10899358 - -
          Affymetrix Rat Gene 1.0 ST Array 10899364 - -
          Affymetrix Rat Gene 1.0 ST Array 10899360 - -
          Affymetrix Rat Gene 1.0 ST Array 10899366 - -
          Affymetrix Rat Gene 1.0 ST Array 10899361 - -
          Affymetrix Rat Gene 1.1 ST Array 10899354 - -
          Rat Genome 230 2.0 Array 1368553_PM_at - -
          Rat Genome 230 2.0 Array 1368553_at - -
          Affymetrix HT Rat Focus Array 1368553_at - -
          Affymetrix HT Rat Focus Array 1368553_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700014484.rn.1 TC0700001864.rn.1 RTA TC
          Rat Expression Set 230 1368553_PM_at - -
          Rat Expression Set 230 1368553_at - -
          Affymetrix Rat Gene 1.0 ST Array 10899354 - -
          Rat Genome U34 Set L36088_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21941172 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358119 - -
          RatRef-12 Expression BeadChip ILMN_57154 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline