Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574568_m1
          See other RHD GE Assays ›
          Gene Symbol:
          Rhd
          Gene Name:
          Rh blood group, D antigen
          Gene Aliases:
          Rh, Rhced
          Chromosome Location:
          Chr.5: 153197416 - 153232009 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574568_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rhd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022505.2 NP_071950.1 4 - 5 696 71
          XM_006239268.2 XP_006239330.1 4 - 5 730 71
          GenBank mRNA AB015191.1 - 4 - 5 661 71
          AF531096.1 - 4 - 5 696 71
          BC127471.1 - 4 - 5 685 8374048 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001666.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rhd

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          ammonium transport
          erythrocyte development
          multicellular organismal iron ion homeostasis
          ammonium transmembrane transport
          ammonium transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251877 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169252 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122436 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10431 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24286 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83205 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12228 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12228 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12228 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12228 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700818521 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76052 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51452 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35205 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38014 - -
          Agilent Rat cDNA Microarray (G4105A) 700818521 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872819 - -
          Affymetrix Rat Gene 1.0 ST Array 10872816 - -
          Affymetrix Rat Gene 1.0 ST Array 10872822 - -
          Affymetrix Rat Gene 1.0 ST Array 10872815 - -
          Affymetrix Rat Gene 1.0 ST Array 10872823 - -
          Affymetrix Rat Gene 1.0 ST Array 10872818 - -
          Affymetrix Rat Gene 1.0 ST Array 10872821 - -
          Affymetrix Rat Gene 1.0 ST Array 10872824 - -
          Affymetrix Rat Gene 1.0 ST Array 10872825 - -
          Affymetrix Rat Gene 1.0 ST Array 10872817 - -
          Affymetrix Rat Gene 1.0 ST Array 10872820 - -
          Affymetrix Rat Gene 1.1 ST Array 10872814 - -
          Rat Genome 230 2.0 Array 1370079_PM_at - -
          Rat Genome 230 2.0 Array 1370079_at - -
          Affymetrix HT Rat Focus Array 1370079_at - -
          Affymetrix HT Rat Focus Array 1370079_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500012017.rn.1 TC0500001666.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012024.rn.1 TC0500001666.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012020.rn.1 TC0500001666.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012022.rn.1 TC0500001666.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012025.rn.1 TC0500001666.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012026.rn.1 TC0500001666.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012023.rn.1 TC0500001666.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012019.rn.1 TC0500001666.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012021.rn.1 TC0500001666.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012028.rn.1 TC0500001666.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012018.rn.1 TC0500001666.rn.1 RTA TC
          Rat Expression Set 230 1370079_PM_at - -
          Rat Expression Set 230 1370079_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872814 - -
          Rat Genome U34 Set AB015191_g_at - -
          Rat Genome U34 Set AB015191_at - -
          Rat Genome U34 Set rc_AA818592_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21615986 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373400 - -
          RatRef-12 Expression BeadChip ILMN_50567 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline