Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574600_m1
          See other MTHFD1 GE Assays ›
          Gene Symbol:
          Mthfd1
          Gene Name:
          methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1, methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthetase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 99282850 - 99350367 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574600_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mthfd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022508.1 NP_071953.1 21 - 22 2447 87
          XM_006240249.2 XP_006240311.1 21 - 22 2586 87
          GenBank mRNA BC089800.1 - 21 - 22 2161 7323085 87
          J05519.1 - 21 - 22 2447 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001008.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mthfd1

          Gene Ontology Categories:

          Function(s) Process(es)

          histidine biosynthetic process
          purine nucleotide biosynthetic process
          one-carbon metabolic process
          methionine biosynthetic process
          nucleotide metabolic process
          folic acid-containing compound biosynthetic process
          tetrahydrofolate interconversion
          tetrahydrofolate metabolic process
          oxidation-reduction process
          formate-tetrahydrofolate ligase activity
          methenyltetrahydrofolate cyclohydrolase activity
          methylenetetrahydrofolate dehydrogenase [NAD(P)+] activity
          methylenetetrahydrofolate dehydrogenase (NADP+) activity
          ATP binding
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P481696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776597 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107085 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88265 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14053 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87931 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5484 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2827 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180617 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125882 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P481696 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P481696 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P481696 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P481696 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700302713 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92908 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26491 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94250 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16200 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101784 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83686 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8290 - -
          Agilent Rat cDNA Microarray (G4105A) 700302713 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10885384 - -
          Affymetrix Rat Gene 1.0 ST Array 10885388 - -
          Affymetrix Rat Gene 1.0 ST Array 10885391 - -
          Affymetrix Rat Gene 1.0 ST Array 10885392 - -
          Affymetrix Rat Gene 1.0 ST Array 10885385 - -
          Affymetrix Rat Gene 1.0 ST Array 10839729 - -
          Affymetrix Rat Gene 1.0 ST Array 10885390 - -
          Affymetrix Rat Gene 1.0 ST Array 10885387 - -
          Affymetrix Rat Gene 1.0 ST Array 10885389 - -
          Affymetrix Rat Gene 1.0 ST Array 10885386 - -
          Affymetrix Rat Gene 1.1 ST Array 10885382 - -
          Affymetrix Rat Gene 1.1 ST Array 10839726 - -
          Rat Genome 230 2.0 Array 1368181_at - -
          Rat Genome 230 2.0 Array 1368181_PM_at - -
          Affymetrix HT Rat Focus Array 1368181_at - -
          Affymetrix HT Rat Focus Array 1368181_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600006981.rn.1 TC0600001008.rn.1 RTA TC
          Rat Expression Set 230 1368181_PM_at - -
          Rat Expression Set 230 1368181_at - -
          Affymetrix Rat Gene 1.0 ST Array 10885382 - -
          Affymetrix Rat Gene 1.0 ST Array 10839726 - -
          Rat Genome U34 Set J05519_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21380029 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365523 - -
          RatRef-12 Expression BeadChip ILMN_53784 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0