Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574615_m1
          See other SMN1 GE Assays ›
          Gene Symbol:
          Smn1
          Gene Name:
          survival of motor neuron 1, telomeric
          Gene Aliases:
          Smn
          Chromosome Location:
          Chr.2: 30360101 - 30371147 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574615_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022509.1 NP_071954.1 1 - 2 115 130
          GenBank mRNA AF044910.1 - 1 - 2 95 130
          AY876898.1 - 1 - 2 85 130
          BC062404.1 - 1 - 2 100 5621519 130
          FQ212707.1 - 1 - 2 111 130
          U75369.1 - 1 - 2 115 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000262.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smn1

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          spliceosomal complex assembly
          spliceosomal snRNP assembly
          mRNA processing
          microtubule depolymerization
          chemical synaptic transmission
          nervous system development
          axonogenesis
          RNA binding
          fibroblast growth factor binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P109834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102850 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18263 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19388 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109834 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109834 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P109834 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P581845 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24128 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63450 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10812745 - -
          Affymetrix Rat Gene 1.0 ST Array 10812739 - -
          Affymetrix Rat Gene 1.0 ST Array 10812743 - -
          Affymetrix Rat Gene 1.0 ST Array 10812746 - -
          Affymetrix Rat Gene 1.0 ST Array 10812747 - -
          Affymetrix Rat Gene 1.0 ST Array 10812741 - -
          Affymetrix Rat Gene 1.0 ST Array 10812740 - -
          Affymetrix Rat Gene 1.0 ST Array 10812742 - -
          Affymetrix Rat Gene 1.0 ST Array 10812744 - -
          Affymetrix Rat Gene 1.0 ST Array 10812738 - -
          Affymetrix Rat Gene 1.1 ST Array 10812737 - -
          Rat Genome 230 2.0 Array 1367883_PM_at - -
          Rat Genome 230 2.0 Array 1367883_at - -
          Affymetrix HT Rat Focus Array 1367883_at - -
          Affymetrix HT Rat Focus Array 1367883_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200001463.rn.1 TC0200000262.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200001462.rn.1 TC0200000262.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200001461.rn.1 TC0200000262.rn.1 RTA TC
          Rat Expression Set 230 1367883_PM_at - -
          Rat Expression Set 230 1367883_at - -
          Affymetrix Rat Gene 1.0 ST Array 10812737 - -
          Rat Genome U34 Set AF044910_g_at - -
          Rat Genome U34 Set AF044910_at - -
          Rat Genome U34 Set AF044910_g_at - -
          Rat Genome U34 Set AF044910_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21716338 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357745 - -
          RatRef-12 Expression BeadChip ILMN_50158 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline