Search
Search

Serpina1| Gene Symbol: Serpina1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022519.2 | NP_071964.2 | 5 - 6 | 1297 | 111 | |
| XM_006240456.1 | XP_006240518.1 | 4 - 5 | 1154 | 111 | ||
| GenBank mRNA | BC078824.1 | - | 5 - 6 | 1297 | 7131535 | 111 |
| D00675.1 | - | 4 - 5 | 1100 | 111 | ||
| FQ209479.1 | - | 3 - 4 | 611 | 111 | ||
| FQ209494.1 | - | 3 - 4 | 614 | 111 | ||
| FQ209517.1 | - | 3 - 4 | 611 | 111 | ||
| FQ209677.1 | - | 3 - 4 | 611 | 111 | ||
| FQ209716.1 | - | 3 - 4 | 612 | 111 | ||
| FQ209779.1 | - | 3 - 4 | 612 | 111 | ||
| FQ210169.1 | - | 3 - 4 | 611 | 111 | ||
| FQ210668.1 | - | 2 - 3 | 233 | 111 | ||
| FQ210685.1 | - | 3 - 4 | 611 | 111 | ||
| FQ210775.1 | - | 3 - 4 | 611 | 111 | ||
| FQ210989.1 | - | 3 - 4 | 612 | 111 | ||
| FQ210996.1 | - | 3 - 4 | 612 | 111 | ||
| FQ211012.1 | - | 3 - 4 | 611 | 111 | ||
| FQ211042.1 | - | 3 - 4 | 611 | 111 | ||
| FQ211203.1 | - | 3 - 4 | 611 | 111 | ||
| FQ211353.1 | - | 3 - 4 | 611 | 111 | ||
| FQ211455.1 | - | 3 - 4 | 477 | 111 | ||
| FQ218252.1 | - | 3 - 4 | 612 | 111 | ||
| FQ218261.1 | - | 3 - 4 | 611 | 111 | ||
| FQ218460.1 | - | 3 - 4 | 611 | 111 | ||
| FQ218555.1 | - | 3 - 4 | 611 | 111 | ||
| FQ218567.1 | - | 3 - 4 | 611 | 111 | ||
| FQ218654.1 | - | 3 - 4 | 611 | 111 | ||
| FQ218694.1 | - | 3 - 4 | 612 | 111 | ||
| FQ218710.1 | - | 3 - 4 | 610 | 111 | ||
| FQ218855.1 | - | 3 - 4 | 611 | 111 | ||
| FQ218857.1 | - | 3 - 4 | 612 | 111 | ||
| FQ218947.1 | - | 3 - 4 | 612 | 111 | ||
| FQ218955.1 | - | 3 - 4 | 612 | 111 | ||
| FQ218993.1 | - | 3 - 4 | 611 | 111 | ||
| FQ219224.1 | - | 3 - 4 | 611 | 111 | ||
| FQ219268.1 | - | 3 - 4 | 611 | 111 | ||
| FQ219276.1 | - | 3 - 4 | 611 | 111 | ||
| FQ219303.1 | - | 3 - 4 | 611 | 111 | ||
| FQ219383.1 | - | 3 - 4 | 611 | 111 | ||
| FQ219441.1 | - | 3 - 4 | 611 | 111 | ||
| FQ219458.1 | - | 3 - 4 | 611 | 111 | ||
| FQ219621.1 | - | 3 - 4 | 611 | 111 | ||
| FQ219652.1 | - | 3 - 4 | 611 | 111 | ||
| FQ219741.1 | - | 4 - 5 | 1091 | 111 | ||
| FQ219765.1 | - | 3 - 4 | 111 | |||
| M32247.1 | - | 3 - 4 | 1037 | 111 | ||
| X16273.1 | - | 3 - 4 | 483 | 111 | ||
Set Membership: |
|