Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574708_m1
          See other DAP GE Assays ›
          Gene Symbol:
          Dap
          Gene Name:
          death-associated protein
          Gene Aliases:
          Rap7a
          Chromosome Location:
          Chr.2: 84275884 - 84328998 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574708_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022526.2 NP_071971.1 2 - 3 343 93
          GenBank mRNA BC060569.1 - 2 - 3 161 6890457 93
          FQ218203.1 - 2 - 3 228 93
          FQ228698.1 - 2 - 3 181 93
          FQ228865.1 - 2 - 3 187 93
          U05334.1 - 2 - 3 344 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000713.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dap

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagy
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of autophagy
          negative regulation of NF-kappaB transcription factor activity
          cellular response to amino acid starvation
          negative regulation of transcription, DNA-templated
          apoptotic signaling pathway
          death domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P099923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38763 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63657 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175281 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56270 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134361 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160875 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96059 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126926 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P099923 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P099923 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P099923 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P306639 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700485510 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29426 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46558 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29425 - -
          Agilent Rat cDNA Microarray (G4105A) 700485510 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10814104 - -
          Affymetrix Rat Gene 1.0 ST Array 10814101 - -
          Affymetrix Rat Gene 1.0 ST Array 10814100 - -
          Affymetrix Rat Gene 1.0 ST Array 10814099 - -
          Affymetrix Rat Gene 1.0 ST Array 10814102 - -
          Affymetrix Rat Gene 1.0 ST Array 10814103 - -
          Affymetrix Rat Gene 1.1 ST Array 10814098 - -
          Rat Genome 230 2.0 Array 1395881_PM_at - -
          Rat Genome 230 2.0 Array 1369941_at - -
          Rat Genome 230 2.0 Array 1395881_at - -
          Rat Genome 230 2.0 Array 1369941_PM_at - -
          Affymetrix HT Rat Focus Array 1369941_PM_at - -
          Affymetrix HT Rat Focus Array 1369941_at - -
          Affymetrix HT Rat Focus Array 1395881_at - -
          Affymetrix HT Rat Focus Array 1395881_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200004396.rn.1 TC0200000713.rn.1 RTA TC
          Rat Expression Set 230 1369941_at - -
          Rat Expression Set 230 1369941_PM_at - -
          Rat Expression Set 230 1395881_PM_at - -
          Rat Expression Set 230 1395881_at - -
          Affymetrix Rat Gene 1.0 ST Array 10814098 - -
          Rat Genome U34 Set rc_AA851650_s_at - -
          Rat Genome U34 Set rc_AI178788_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22389695 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362377 - -
          RatRef-12 Expression BeadChip ILMN_64501 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline