Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574758_m1
          See other TCN2 GE Assays ›
          Gene Symbol:
          Tcn2
          Gene Name:
          transcobalamin 2
          Gene Aliases:
          Tc2, Tcn2p
          Chromosome Location:
          Chr.14: 84174139 - 84189299 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574758_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022534.1 NP_071979.1 7 - 8 1155 108
          XM_006251366.2 XP_006251428.1 8 - 9 1346 108
          XM_006251367.2 XP_006251429.1 8 - 9 1340 108
          XM_006251368.2 XP_006251430.1 8 - 9 1285 108
          XM_006251370.2 XP_006251432.1 8 - 9 1278 108
          GenBank mRNA AF054810.1 - 7 - 8 1155 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002141.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tcn2

          Gene Ontology Categories:

          Function(s) Process(es)

          cobalt ion transport
          cobalamin metabolic process
          cobalamin transport
          cobalamin binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P544351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91086 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70507 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137697 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P544351 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P544351 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P544351 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P544351 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700330269 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33646 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36622 - -
          Agilent Rat cDNA Microarray (G4105A) 700330269 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10778030 - -
          Affymetrix Rat Gene 1.0 ST Array 10778033 - -
          Affymetrix Rat Gene 1.0 ST Array 10778029 - -
          Affymetrix Rat Gene 1.0 ST Array 10778031 - -
          Affymetrix Rat Gene 1.0 ST Array 10778034 - -
          Affymetrix Rat Gene 1.0 ST Array 10778032 - -
          Affymetrix Rat Gene 1.0 ST Array 10778036 - -
          Affymetrix Rat Gene 1.0 ST Array 10778035 - -
          Affymetrix Rat Gene 1.0 ST Array 10778037 - -
          Affymetrix Rat Gene 1.1 ST Array 10778028 - -
          Rat Genome 230 2.0 Array 1367765_at - -
          Rat Genome 230 2.0 Array 1367765_PM_at - -
          Affymetrix HT Rat Focus Array 1367765_PM_at - -
          Affymetrix HT Rat Focus Array 1367765_at - -
          Affymetrix Rat Clariom S Assay PSR1400015735.rn.1 TC1400002141.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400015736.rn.1 TC1400002141.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400015738.rn.1 TC1400002141.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400015737.rn.1 TC1400002141.rn.1 RTA TC
          Rat Expression Set 230 1367765_PM_at - -
          Rat Expression Set 230 1367765_at - -
          Affymetrix Rat Gene 1.0 ST Array 10778028 - -
          Rat Genome U34 Set rc_AA893702_s_at - -
          Rat Genome U34 Set rc_AA819899_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21354815 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355714 - -
          RatRef-12 Expression BeadChip ILMN_64496 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline