Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574804_s1
          See other RHOB GE Assays ›
          Gene Symbol:
          Rhob
          Gene Name:
          ras homolog family member B
          Gene Aliases:
          Arhb
          Chromosome Location:
          Chr.6: 33689127 - 33691301 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574804_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rhob
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022542.1 NP_071987.1 1 452 75
          GenBank mRNA M74295.1 - 1 452 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rhob

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          intracellular protein transport
          obsolete transformed cell apoptotic process
          cell adhesion
          small GTPase mediated signal transduction
          endosome to lysosome transport
          cell differentiation
          positive regulation of apoptotic process
          positive regulation of angiogenesis
          negative regulation of cell cycle
          cellular response to hydrogen peroxide
          cellular response to ionizing radiation
          GTP binding
          GDP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P141727 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49891 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141727 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12245 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12245 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141727 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P141727 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12245 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12245 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700313882 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34744 - -
          Agilent Rat cDNA Microarray (G4105A) 700313882 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10889179 - -
          Affymetrix Rat Gene 1.0 ST Array 10889180 - -
          Affymetrix Rat Gene 1.0 ST Array 10889178 - -
          Affymetrix Rat Gene 1.1 ST Array 10889177 - -
          Rat Genome 230 2.0 Array 1369958_at - -
          Rat Genome 230 2.0 Array 1369958_PM_at - -
          Affymetrix HT Rat Focus Array 1369958_PM_at - -
          Affymetrix HT Rat Focus Array 1369958_at - -
          Affymetrix Rat Clariom S Assay PSR0600015447.rn.1 TC0600002109.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600015446.rn.1 TC0600002109.rn.1 RTA TC
          Rat Expression Set 230 1369958_at - -
          Rat Expression Set 230 1369958_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10889177 - -
          Rat Genome U34 Set rc_AA900505_at - -
          Rat Genome U34 Set rc_AA998352_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21275762 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351285 - -
          RatRef-12 Expression BeadChip ILMN_58180 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0