Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574868_m1
          See other TXNRD2 GE Assays ›
          Gene Symbol:
          Txnrd2
          Gene Name:
          thioredoxin reductase 2
          Gene Aliases:
          Tr3, Trxr2, Trxrd2
          Chromosome Location:
          Chr.11: 86667994 - 86716063 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574868_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Txnrd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022584.2 NP_072106.1 2 - 3 183 58
          GenBank mRNA AF072865.1 - 2 - 3 204 58
          BC085734.1 - 2 - 3 183 7191992 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001984.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Txnrd2

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to oxygen radical
          heart development
          response to selenium ion
          hemopoiesis
          response to drug
          response to hydrogen peroxide
          cell redox homeostasis
          detoxification of mercury ion
          response to hyperoxia
          oxidation-reduction process
          thioredoxin-disulfide reductase activity
          mercury (II) reductase activity
          protein homodimerization activity
          mercury ion binding
          protein dimerization activity
          flavin adenine dinucleotide binding
          NADP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P160658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218095 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125618 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84385 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151693 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88872 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99525 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160658 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160658 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P160658 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P252210 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700695176 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701829326 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27620 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79269 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76964 - -
          Agilent Rat cDNA Microarray (G4105A) 700695176 - -
          Agilent Rat cDNA Microarray (G4105A) 701829326 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10755571 - -
          Affymetrix Rat Gene 1.0 ST Array 10755561 - -
          Affymetrix Rat Gene 1.0 ST Array 10755563 - -
          Affymetrix Rat Gene 1.0 ST Array 10755578 - -
          Affymetrix Rat Gene 1.0 ST Array 10755572 - -
          Affymetrix Rat Gene 1.0 ST Array 10755564 - -
          Affymetrix Rat Gene 1.0 ST Array 10755570 - -
          Affymetrix Rat Gene 1.0 ST Array 10755566 - -
          Affymetrix Rat Gene 1.0 ST Array 10755576 - -
          Affymetrix Rat Gene 1.0 ST Array 10755577 - -
          Affymetrix Rat Gene 1.0 ST Array 10755579 - -
          Affymetrix Rat Gene 1.0 ST Array 10755568 - -
          Affymetrix Rat Gene 1.0 ST Array 10755565 - -
          Affymetrix Rat Gene 1.0 ST Array 10755574 - -
          Affymetrix Rat Gene 1.0 ST Array 10755575 - -
          Affymetrix Rat Gene 1.0 ST Array 10755567 - -
          Affymetrix Rat Gene 1.0 ST Array 10755562 - -
          Affymetrix Rat Gene 1.0 ST Array 10755573 - -
          Affymetrix Rat Gene 1.1 ST Array 10755560 - -
          Rat Genome 230 2.0 Array 1368309_at - -
          Rat Genome 230 2.0 Array 1368309_PM_at - -
          Affymetrix HT Rat Focus Array 1368309_PM_at - -
          Affymetrix HT Rat Focus Array 1368309_at - -
          Affymetrix Rat Clariom S Assay PSR1100013012.rn.1 TC1100001984.rn.1 RTA TC
          Rat Expression Set 230 1368309_PM_at - -
          Rat Expression Set 230 1368309_at - -
          Affymetrix Rat Gene 1.0 ST Array 10755560 - -
          Rat Genome U34 Set AF072865_g_at - -
          Rat Genome U34 Set AF072865_at - -
          Rat Genome U34 Set AF072865_g_at - -
          Rat Genome U34 Set AF072865_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21467109 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374002 - -
          RatRef-12 Expression BeadChip ILMN_70004 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline