Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574968_m1
          See other TEP1 GE Assays ›
          Gene Symbol:
          Tep1
          Gene Name:
          telomerase associated protein 1
          Gene Aliases:
          Tlp1
          Chromosome Location:
          Chr.15: 27751186 - 27798839 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574968_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tep1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022591.2 NP_072113.1 51 - 52 7546 65
          GenBank mRNA U89282.1 - 51 - 52 7547 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001555.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tep1

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance via recombination
          RNA-dependent DNA biosynthetic process
          telomere maintenance via telomerase
          telomerase activity
          RNA binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P033569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935522 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166971 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71189 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85231 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144343 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116354 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47015 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107466 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033569 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033569 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P033569 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P544167 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701932996 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700885027 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702149780 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83917 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95190 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96388 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35881 - -
          Agilent Rat cDNA Microarray (G4105A) 701932996 - -
          Agilent Rat cDNA Microarray (G4105A) 700885027 - -
          Agilent Rat cDNA Microarray (G4105A) 702149780 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783092 - -
          Affymetrix Rat Gene 1.0 ST Array 10783105 - -
          Affymetrix Rat Gene 1.0 ST Array 10783122 - -
          Affymetrix Rat Gene 1.0 ST Array 10783138 - -
          Affymetrix Rat Gene 1.0 ST Array 10783108 - -
          Affymetrix Rat Gene 1.0 ST Array 10783129 - -
          Affymetrix Rat Gene 1.0 ST Array 10783115 - -
          Affymetrix Rat Gene 1.0 ST Array 10783111 - -
          Affymetrix Rat Gene 1.0 ST Array 10783087 - -
          Affymetrix Rat Gene 1.0 ST Array 10783128 - -
          Affymetrix Rat Gene 1.0 ST Array 10783139 - -
          Affymetrix Rat Gene 1.0 ST Array 10783118 - -
          Affymetrix Rat Gene 1.0 ST Array 10783096 - -
          Affymetrix Rat Gene 1.0 ST Array 10783123 - -
          Affymetrix Rat Gene 1.0 ST Array 10783110 - -
          Affymetrix Rat Gene 1.0 ST Array 10783107 - -
          Affymetrix Rat Gene 1.0 ST Array 10783114 - -
          Affymetrix Rat Gene 1.0 ST Array 10783130 - -
          Affymetrix Rat Gene 1.0 ST Array 10783136 - -
          Affymetrix Rat Gene 1.0 ST Array 10783116 - -
          Affymetrix Rat Gene 1.0 ST Array 10783125 - -
          Affymetrix Rat Gene 1.0 ST Array 10783112 - -
          Affymetrix Rat Gene 1.0 ST Array 10783124 - -
          Affymetrix Rat Gene 1.0 ST Array 10783135 - -
          Affymetrix Rat Gene 1.0 ST Array 10783113 - -
          Affymetrix Rat Gene 1.0 ST Array 10783137 - -
          Affymetrix Rat Gene 1.0 ST Array 10783093 - -
          Affymetrix Rat Gene 1.0 ST Array 10783132 - -
          Affymetrix Rat Gene 1.0 ST Array 10783085 - -
          Affymetrix Rat Gene 1.0 ST Array 10783097 - -
          Affymetrix Rat Gene 1.0 ST Array 10783088 - -
          Affymetrix Rat Gene 1.0 ST Array 10783101 - -
          Affymetrix Rat Gene 1.0 ST Array 10783086 - -
          Affymetrix Rat Gene 1.0 ST Array 10783133 - -
          Affymetrix Rat Gene 1.0 ST Array 10783089 - -
          Affymetrix Rat Gene 1.0 ST Array 10783127 - -
          Affymetrix Rat Gene 1.0 ST Array 10783109 - -
          Affymetrix Rat Gene 1.0 ST Array 10783131 - -
          Affymetrix Rat Gene 1.0 ST Array 10783134 - -
          Affymetrix Rat Gene 1.0 ST Array 10783117 - -
          Affymetrix Rat Gene 1.0 ST Array 10783098 - -
          Affymetrix Rat Gene 1.0 ST Array 10783095 - -
          Affymetrix Rat Gene 1.0 ST Array 10783094 - -
          Affymetrix Rat Gene 1.0 ST Array 10783120 - -
          Affymetrix Rat Gene 1.0 ST Array 10783091 - -
          Affymetrix Rat Gene 1.0 ST Array 10783121 - -
          Affymetrix Rat Gene 1.0 ST Array 10783102 - -
          Affymetrix Rat Gene 1.0 ST Array 10783104 - -
          Affymetrix Rat Gene 1.0 ST Array 10783103 - -
          Affymetrix Rat Gene 1.0 ST Array 10783084 - -
          Affymetrix Rat Gene 1.0 ST Array 10783099 - -
          Affymetrix Rat Gene 1.0 ST Array 10783126 - -
          Affymetrix Rat Gene 1.0 ST Array 10783106 - -
          Affymetrix Rat Gene 1.0 ST Array 10783119 - -
          Affymetrix Rat Gene 1.0 ST Array 10783100 - -
          Affymetrix Rat Gene 1.0 ST Array 10783090 - -
          Affymetrix Rat Gene 1.1 ST Array 10783083 - -
          Rat Genome 230 2.0 Array 1368012_at - -
          Rat Genome 230 2.0 Array 1368012_PM_at - -
          Affymetrix HT Rat Focus Array 1368012_at - -
          Affymetrix HT Rat Focus Array 1368012_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500009777.rn.1 TC1500001555.rn.1 RTA TC
          Rat Expression Set 230 1368012_PM_at - -
          Rat Expression Set 230 1368012_at - -
          Affymetrix Rat Gene 1.0 ST Array 10783083 - -
          Rat Genome U34 Set U89282_at - -
          Rat Genome U34 Set H33937_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20752295 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353251 - -
          RatRef-12 Expression BeadChip ILMN_49889 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline