Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574992_m1
          See other ECH1 GE Assays ›
          Gene Symbol:
          Ech1
          Gene Name:
          enoyl CoA hydratase 1, peroxisomal
          Gene Aliases:
          Pxel
          Chromosome Location:
          Chr.1: 87009798 - 87015996 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574992_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ech1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022594.1 NP_072116.1 3 - 4 351 64
          GenBank mRNA BC060565.1 - 3 - 4 357 64
          BC062226.1 - 3 - 4 357 6890529 64
          FQ214121.1 - 3 - 4 353 64
          FQ230435.1 - 3 - 4 364 64
          U08976.1 - 3 - 4 351 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001192.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ech1

          Panther Classification:

          Molecular Function -

          hydratase lyase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid beta-oxidation
          receptor binding
          isomerase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P241885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540921 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171048 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P241885 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P241885 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P241885 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P241885 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700247611 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8070 - -
          Agilent Rat cDNA Microarray (G4105A) 700247611 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10705638 - -
          Affymetrix Rat Gene 1.0 ST Array 10705635 - -
          Affymetrix Rat Gene 1.0 ST Array 10705632 - -
          Affymetrix Rat Gene 1.0 ST Array 10705634 - -
          Affymetrix Rat Gene 1.0 ST Array 10705633 - -
          Affymetrix Rat Gene 1.0 ST Array 10705637 - -
          Affymetrix Rat Gene 1.0 ST Array 10705640 - -
          Affymetrix Rat Gene 1.0 ST Array 10705639 - -
          Affymetrix Rat Gene 1.0 ST Array 10705641 - -
          Affymetrix Rat Gene 1.1 ST Array 10705631 - -
          Rat Genome 230 2.0 Array 1386885_PM_at - -
          Rat Genome 230 2.0 Array 1386885_at - -
          Affymetrix HT Rat Focus Array 1386885_at - -
          Affymetrix HT Rat Focus Array 1386885_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100009021.rn.1 TC0100001192.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100009020.rn.1 TC0100001192.rn.1 RTA TC
          Rat Expression Set 230 1386885_at - -
          Rat Expression Set 230 1386885_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10705631 - -
          Rat Genome U34 Set U08976_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21593647 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364554 - -
          RatRef-12 Expression BeadChip ILMN_68003 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline