Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575067_m1
          See other PIM3 GE Assays ›
          Gene Symbol:
          Pim3
          Gene Name:
          Pim-3 proto-oncogene, serine/threonine kinase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 129856467 - 129863441 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575067_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pim3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022602.1 NP_072124.1 804 75
          GenBank mRNA AF057026.1 - 804 75
          AF086624.1 - 1006 75
          BC097317.1 - 783 7446368 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001627.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pim3

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          apoptotic process
          regulation of mitotic cell cycle
          histone phosphorylation
          negative regulation of apoptotic process
          protein autophosphorylation
          negative regulation of insulin secretion involved in cellular response to glucose stimulus
          protein serine/threonine kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1034541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151898 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1034541 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1034541 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1034541 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1034541 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10898477 - -
          Affymetrix Rat Gene 1.0 ST Array 10898480 - -
          Affymetrix Rat Gene 1.0 ST Array 10898478 - -
          Affymetrix Rat Gene 1.0 ST Array 10898475 - -
          Affymetrix Rat Gene 1.0 ST Array 10898479 - -
          Affymetrix Rat Gene 1.0 ST Array 10898476 - -
          Affymetrix Rat Gene 1.1 ST Array 10898474 - -
          Rat Genome 230 2.0 Array 1367725_PM_at - -
          Rat Genome 230 2.0 Array 1367725_at - -
          Affymetrix HT Rat Focus Array 1367725_at - -
          Affymetrix HT Rat Focus Array 1367725_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700012438.rn.1 TC0700001627.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700012439.rn.1 TC0700001627.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700012432.rn.1 TC0700001627.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700012434.rn.1 TC0700001627.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700012437.rn.1 TC0700001627.rn.1 RTA TC
          Rat Expression Set 230 1367725_at - -
          Rat Expression Set 230 1367725_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10898474 - -
          Rat Genome U34 Set AF086624_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21426517 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360660 - -
          RatRef-12 Expression BeadChip ILMN_60058 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline