Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575362_m1
          See other ONECUT1 GE Assays ›
          Gene Symbol:
          Onecut1
          Gene Name:
          one cut homeobox 1
          Gene Aliases:
          Hnf6
          Chromosome Location:
          Chr.8: 81765860 - 81799894 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575362_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Onecut1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022671.2 NP_073162.1 1 - 2 1337 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001105.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Onecut1

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          regulation of cell-matrix adhesion
          epithelial cell development
          glucose metabolic process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          endoderm development
          anatomical structure morphogenesis
          cell differentiation
          B cell differentiation
          positive regulation of cell migration
          negative regulation of transforming growth factor beta receptor signaling pathway
          pancreas development
          endocrine pancreas development
          cilium assembly
          cell fate commitment
          positive regulation of transcription from RNA polymerase II promoter
          spleen development
          system development
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P120342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943928 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175839 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44225 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120342 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120342 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P120342 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P701060 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86574 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93475 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19568 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10911572 - -
          Affymetrix Rat Gene 1.0 ST Array 10911571 - -
          Affymetrix Rat Gene 1.0 ST Array 10911574 - -
          Affymetrix Rat Gene 1.0 ST Array 10911575 - -
          Affymetrix Rat Gene 1.0 ST Array 10911573 - -
          Affymetrix Rat Gene 1.1 ST Array 10911570 - -
          Rat Genome 230 2.0 Array 1371034_PM_at - -
          Rat Genome 230 2.0 Array 1387760_PM_a_at - -
          Rat Genome 230 2.0 Array 1371034_at - -
          Rat Genome 230 2.0 Array 1387760_a_at - -
          Affymetrix HT Rat Focus Array 1387760_a_at - -
          Affymetrix HT Rat Focus Array 1387760_PM_a_at - -
          Affymetrix HT Rat Focus Array 1371034_at - -
          Affymetrix HT Rat Focus Array 1371034_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800008032.rn.1 TC0800001105.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800008034.rn.1 TC0800001105.rn.1 RTA TC
          Rat Expression Set 230 1371034_PM_at - -
          Rat Expression Set 230 1387760_PM_a_at - -
          Rat Expression Set 230 1371034_at - -
          Rat Expression Set 230 1387760_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10911570 - -
          Rat Genome U34 Set Y14933mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20711618 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363497 - -
          RatRef-12 Expression BeadChip ILMN_54767 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline