Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575374_m1
          See other PPP1R1A GE Assays ›
          Gene Symbol:
          Ppp1r1a
          Gene Name:
          protein phosphatase 1, regulatory (inhibitor) subunit 1A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 145146480 - 145154146 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575374_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp1r1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022676.3 NP_073167.1 5 - 6 536 64
          XM_006242437.2 XP_006242499.1 5 - 6 548 64
          GenBank mRNA AJ276593.1 - 5 - 6 409 64
          AY648296.1 - 5 - 6 536 64
          BC078820.1 - 5 - 6 537 7127505 64
          FQ217772.1 - 5 - 6 505 64
          J05592.1 - 5 - 6 408 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700004006.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp1r1a

          Panther Classification:

          Molecular Function -

          phosphatase inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          glycogen metabolic process
          negative regulation of protein kinase activity
          intracellular signal transduction
          protein serine/threonine phosphatase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P145261 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962380 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P145261 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12287 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12287 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P145261 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P145261 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12287 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12287 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701716678 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700280368 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102379 - -
          Agilent Rat cDNA Microarray (G4105A) 701716678 - -
          Agilent Rat cDNA Microarray (G4105A) 700280368 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10907740 - -
          Affymetrix Rat Gene 1.0 ST Array 10907739 - -
          Affymetrix Rat Gene 1.0 ST Array 10907747 - -
          Affymetrix Rat Gene 1.0 ST Array 10907741 - -
          Affymetrix Rat Gene 1.0 ST Array 10907742 - -
          Affymetrix Rat Gene 1.0 ST Array 10907744 - -
          Affymetrix Rat Gene 1.0 ST Array 10907746 - -
          Affymetrix Rat Gene 1.0 ST Array 10907743 - -
          Affymetrix Rat Gene 1.0 ST Array 10907745 - -
          Affymetrix Rat Gene 1.0 ST Array 10907748 - -
          Affymetrix Rat Gene 1.1 ST Array 10907738 - -
          Rat Genome 230 2.0 Array 1386968_at - -
          Rat Genome 230 2.0 Array 1384203_PM_at - -
          Rat Genome 230 2.0 Array 1383191_at - -
          Rat Genome 230 2.0 Array 1386968_PM_at - -
          Rat Genome 230 2.0 Array 1384203_at - -
          Rat Genome 230 2.0 Array 1383191_PM_at - -
          Affymetrix HT Rat Focus Array 1383191_at - -
          Affymetrix HT Rat Focus Array 1386968_at - -
          Affymetrix HT Rat Focus Array 1386968_PM_at - -
          Affymetrix HT Rat Focus Array 1383191_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700032454.rn.1 TC0700004006.rn.1 RTA TC
          Rat Expression Set 230 1386968_at - -
          Rat Expression Set 230 1386968_PM_at - -
          Rat Expression Set 230 1383191_PM_at - -
          Rat Expression Set 230 1383191_at - -
          Affymetrix Rat Gene 1.0 ST Array 10907738 - -
          Rat Genome U34 Set J05592_at - -
          Rat Genome U34 Set J05592_g_at - -
          Rat Genome U34 Set rc_AI113332_at - -
          Rat Genome U34 Set rc_AI145995_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21720242 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356921 - -
          RatRef-12 Expression BeadChip ILMN_59675 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline