Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575428_m1
          See other CUL5 GE Assays ›
          Gene Symbol:
          Cul5
          Gene Name:
          cullin 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 58199992 - 58254195 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575428_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cul5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022683.1 NP_073174.1 11 - 12 1318 121
          XM_006243090.2 XP_006243152.1 10 - 11 2031 121
          GenBank mRNA AF135115.1 - 11 - 12 1318 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002563.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cul5

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          ubiquitin-dependent protein catabolic process
          response to osmotic stress
          G-protein coupled receptor signaling pathway
          protein ubiquitination
          cerebral cortex radially oriented cell migration
          radial glia guided migration of Purkinje cell
          regulation of cytosolic calcium ion concentration
          ubiquitin-protein transferase activity
          vasopressin receptor activity
          ubiquitin protein ligase binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P525878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22445 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82595 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159332 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150514 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164852 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158386 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62957 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P525878 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P525878 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P525878 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P525878 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69396 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24070 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7322 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10917482 - -
          Affymetrix Rat Gene 1.0 ST Array 10917473 - -
          Affymetrix Rat Gene 1.0 ST Array 10917481 - -
          Affymetrix Rat Gene 1.0 ST Array 10917466 - -
          Affymetrix Rat Gene 1.0 ST Array 10917463 - -
          Affymetrix Rat Gene 1.0 ST Array 10917468 - -
          Affymetrix Rat Gene 1.0 ST Array 10917465 - -
          Affymetrix Rat Gene 1.0 ST Array 10917464 - -
          Affymetrix Rat Gene 1.0 ST Array 10917472 - -
          Affymetrix Rat Gene 1.0 ST Array 10917478 - -
          Affymetrix Rat Gene 1.0 ST Array 10917480 - -
          Affymetrix Rat Gene 1.0 ST Array 10917469 - -
          Affymetrix Rat Gene 1.0 ST Array 10917479 - -
          Affymetrix Rat Gene 1.0 ST Array 10917467 - -
          Affymetrix Rat Gene 1.0 ST Array 10917474 - -
          Affymetrix Rat Gene 1.0 ST Array 10917470 - -
          Affymetrix Rat Gene 1.0 ST Array 10917471 - -
          Affymetrix Rat Gene 1.0 ST Array 10917475 - -
          Affymetrix Rat Gene 1.0 ST Array 10917477 - -
          Affymetrix Rat Gene 1.1 ST Array 10917462 - -
          Rat Genome 230 2.0 Array 1393930_PM_at - -
          Rat Genome 230 2.0 Array 1396018_PM_at - -
          Rat Genome 230 2.0 Array 1393930_at - -
          Rat Genome 230 2.0 Array 1369068_at - -
          Rat Genome 230 2.0 Array 1396018_at - -
          Rat Genome 230 2.0 Array 1393306_at - -
          Rat Genome 230 2.0 Array 1393306_PM_at - -
          Rat Genome 230 2.0 Array 1369068_PM_at - -
          Affymetrix HT Rat Focus Array 1369068_at - -
          Affymetrix HT Rat Focus Array 1393306_PM_at - -
          Affymetrix HT Rat Focus Array 1369068_PM_at - -
          Affymetrix HT Rat Focus Array 1393306_at - -
          Affymetrix Rat Clariom S Assay PSR0800020690.rn.1 TC0800002563.rn.1 RTA TC
          Rat Expression Set 230 1369068_PM_at - -
          Rat Expression Set 230 1369068_at - -
          Rat Expression Set 230 1393306_PM_at - -
          Rat Expression Set 230 1393306_at - -
          Affymetrix Rat Gene 1.0 ST Array 10917462 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22203961 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369400 - -
          RatRef-12 Expression BeadChip ILMN_66271 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0