Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575535_m1
          See other FLOT1 GE Assays ›
          Gene Symbol:
          Flot1
          Gene Name:
          flotillin 1
          Gene Aliases:
          Rareg
          Chromosome Location:
          Chr.20: 3427890 - 3438409 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575535_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Flot1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022701.2 NP_073192.2 8 - 9 897 99
          XM_006256024.2 XP_006256086.1 8 - 9 916 99
          GenBank mRNA BC064652.1 - 8 - 9 896 5598180 99
          DY569467.1 - 5 - 6 579 99
          U60976.1 - 8 - 9 827 99
          U60977.1 - 7 - 8 901 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001039.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Flot1

          Gene Ontology Categories:

          Function(s) Process(es)

          axonogenesis
          extracellular matrix disassembly
          regulation of Rho protein signal transduction
          protein localization to plasma membrane
          establishment of protein localization to plasma membrane
          protease binding
          protein binding
          ionotropic glutamate receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1023498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835584 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165361 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1023498 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1023498 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1023498 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1023498 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56320 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10831066 - -
          Affymetrix Rat Gene 1.0 ST Array 10831075 - -
          Affymetrix Rat Gene 1.0 ST Array 10831076 - -
          Affymetrix Rat Gene 1.0 ST Array 10831071 - -
          Affymetrix Rat Gene 1.0 ST Array 10831069 - -
          Affymetrix Rat Gene 1.0 ST Array 10831068 - -
          Affymetrix Rat Gene 1.0 ST Array 10831067 - -
          Affymetrix Rat Gene 1.0 ST Array 10831074 - -
          Affymetrix Rat Gene 1.0 ST Array 10831064 - -
          Affymetrix Rat Gene 1.0 ST Array 10831062 - -
          Affymetrix Rat Gene 1.0 ST Array 10831072 - -
          Affymetrix Rat Gene 1.0 ST Array 10831065 - -
          Affymetrix Rat Gene 1.0 ST Array 10831070 - -
          Affymetrix Rat Gene 1.0 ST Array 10831073 - -
          Affymetrix Rat Gene 1.0 ST Array 10831061 - -
          Affymetrix Rat Gene 1.0 ST Array 10831063 - -
          Affymetrix Rat Gene 1.1 ST Array 10831060 - -
          Rat Genome 230 2.0 Array 1368241_a_at - -
          Rat Genome 230 2.0 Array 1368241_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368241_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368241_a_at - -
          Affymetrix Rat Clariom S Assay PSR2000008416.rn.1 TC2000001039.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000008417.rn.1 TC2000001039.rn.1 RTA TC
          Rat Expression Set 230 1368241_PM_a_at - -
          Rat Expression Set 230 1368241_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10831060 - -
          Rat Genome U34 Set U60976_s_at - -
          Rat Genome U34 Set U60977_i_at - -
          Rat Genome U34 Set rc_AI178886_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21271755 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348870 - -
          RatRef-12 Expression BeadChip ILMN_48074 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline