Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575578_m1
          See other PIP GE Assays ›
          Gene Symbol:
          Pip
          Gene Name:
          prolactin induced protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 71320859 - 71325153 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575578_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022708.2 NP_073199.1 2 - 3 218 118
          GenBank mRNA AF054270.1 - 2 - 3 218 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000743.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pip

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          retina homeostasis
          regulation of immune system process
          proteolysis
          positive regulation of gene expression
          negative regulation of T cell apoptotic process
          glycoprotein binding
          aspartic-type endopeptidase activity
          IgG binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P096891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583334 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89981 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P096891 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P096891 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P096891 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12308 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92330 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10854989 - -
          Affymetrix Rat Gene 1.0 ST Array 10854992 - -
          Affymetrix Rat Gene 1.0 ST Array 10854990 - -
          Affymetrix Rat Gene 1.0 ST Array 10854991 - -
          Affymetrix Rat Gene 1.1 ST Array 10854988 - -
          Rat Genome 230 2.0 Array 1367781_at - -
          Rat Genome 230 2.0 Array 1367781_PM_at - -
          Affymetrix HT Rat Focus Array 1367781_PM_at - -
          Affymetrix HT Rat Focus Array 1367781_at - -
          Affymetrix Rat Clariom S Assay PSR0400005480.rn.1 TC0400000743.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400005479.rn.1 TC0400000743.rn.1 RTA TC
          Rat Expression Set 230 1367781_PM_at - -
          Rat Expression Set 230 1367781_at - -
          Affymetrix Rat Gene 1.0 ST Array 10854988 - -
          Rat Genome U34 Set AF054270_g_at - -
          Rat Genome U34 Set AF054270_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20909068 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358925 - -
          RatRef-12 Expression BeadChip ILMN_59680 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline