Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575815_m1
          See other UNC13B GE Assays ›
          Gene Symbol:
          Unc13b
          Gene Name:
          unc-13 homolog B (C. elegans)
          Gene Aliases:
          Munc13-2, Unc13h2
          Chromosome Location:
          Chr.5: 58505449 - 58714396 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575815_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Unc13b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042579.1 NP_001036044.1 26 - 27 2986 68
          NM_022862.1 NP_074053.1 17 - 18 4460 68
          GenBank mRNA AF159706.1 - 26 - 27 2986 68
          U24071.1 - 17 - 18 4460 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000519.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Unc13b

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          neuromuscular junction development
          positive regulation of synaptic vesicle priming
          synaptic vesicle docking
          synaptic vesicle priming
          synaptic transmission, glutamatergic
          intracellular signal transduction
          regulation of short-term neuronal synaptic plasticity
          innervation
          positive regulation of inhibitory postsynaptic potential
          protein binding
          diacylglycerol binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P036011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702800 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56969 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131681 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50045 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90647 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93748 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145981 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83119 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15222 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167375 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176982 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107203 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7937 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119841 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139323 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37803 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116848 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74234 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59334 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58041 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36464 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 419 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 401 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19170 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47722 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102199 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10305 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59999 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83401 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82723 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61896 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036011 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036011 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P036011 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P539738 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21633 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80534 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62417 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70656 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22017 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30281 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3730 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39925 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44118 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71571 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70857 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80388 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11937 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1627 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15560 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82884 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10868447 - -
          Affymetrix Rat Gene 1.0 ST Array 10868458 - -
          Affymetrix Rat Gene 1.0 ST Array 10868466 - -
          Affymetrix Rat Gene 1.0 ST Array 10868476 - -
          Affymetrix Rat Gene 1.0 ST Array 10868479 - -
          Affymetrix Rat Gene 1.0 ST Array 10868478 - -
          Affymetrix Rat Gene 1.0 ST Array 10868462 - -
          Affymetrix Rat Gene 1.0 ST Array 10868456 - -
          Affymetrix Rat Gene 1.0 ST Array 10868468 - -
          Affymetrix Rat Gene 1.0 ST Array 10868439 - -
          Affymetrix Rat Gene 1.0 ST Array 10868451 - -
          Affymetrix Rat Gene 1.0 ST Array 10868459 - -
          Affymetrix Rat Gene 1.0 ST Array 10868457 - -
          Affymetrix Rat Gene 1.0 ST Array 10868453 - -
          Affymetrix Rat Gene 1.0 ST Array 10868463 - -
          Affymetrix Rat Gene 1.0 ST Array 10868449 - -
          Affymetrix Rat Gene 1.0 ST Array 10868461 - -
          Affymetrix Rat Gene 1.0 ST Array 10868472 - -
          Affymetrix Rat Gene 1.0 ST Array 10868470 - -
          Affymetrix Rat Gene 1.0 ST Array 10868477 - -
          Affymetrix Rat Gene 1.0 ST Array 10868480 - -
          Affymetrix Rat Gene 1.0 ST Array 10868460 - -
          Affymetrix Rat Gene 1.0 ST Array 10868474 - -
          Affymetrix Rat Gene 1.0 ST Array 10868443 - -
          Affymetrix Rat Gene 1.0 ST Array 10868444 - -
          Affymetrix Rat Gene 1.0 ST Array 10868445 - -
          Affymetrix Rat Gene 1.0 ST Array 10868467 - -
          Affymetrix Rat Gene 1.0 ST Array 10868473 - -
          Affymetrix Rat Gene 1.0 ST Array 10868448 - -
          Affymetrix Rat Gene 1.0 ST Array 10868475 - -
          Affymetrix Rat Gene 1.0 ST Array 10868438 - -
          Affymetrix Rat Gene 1.0 ST Array 10868469 - -
          Affymetrix Rat Gene 1.0 ST Array 10868446 - -
          Affymetrix Rat Gene 1.0 ST Array 10868464 - -
          Affymetrix Rat Gene 1.0 ST Array 10868455 - -
          Affymetrix Rat Gene 1.0 ST Array 10868471 - -
          Affymetrix Rat Gene 1.0 ST Array 10868454 - -
          Affymetrix Rat Gene 1.0 ST Array 10868450 - -
          Affymetrix Rat Gene 1.0 ST Array 10868465 - -
          Affymetrix Rat Gene 1.0 ST Array 10868452 - -
          Affymetrix Rat Gene 1.0 ST Array 10868442 - -
          Affymetrix Rat Gene 1.1 ST Array 10868437 - -
          Rat Genome 230 2.0 Array 1369331_a_at - -
          Rat Genome 230 2.0 Array 1369331_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369331_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369331_a_at - -
          Affymetrix Rat Clariom S Assay PSR0500003206.rn.1 TC0500000519.rn.1 RTA TC
          Rat Expression Set 230 1369331_PM_a_at - -
          Rat Expression Set 230 1369331_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10868437 - -
          Rat Genome U34 Set U24071_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21569284 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348784 - -
          RatRef-12 Expression BeadChip ILMN_58626 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline