Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575971_m1
          See other GALNT7 GE Assays ›
          Gene Symbol:
          Galnt7
          Gene Name:
          polypeptide N-acetylgalactosaminyltransferase 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 35935035 - 36062734 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575971_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Galnt7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022926.1 NP_075215.1 9 - 10 1664 70
          XM_006253083.1 XP_006253145.1 9 - 10 1689 70
          GenBank mRNA AF076167.1 - 9 - 10 1664 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000455.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Galnt7

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein O-linked glycosylation
          biological_process
          molecular_function
          polypeptide N-acetylgalactosaminyltransferase activity
          carbohydrate binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P406169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110381 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139304 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 530 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73750 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8390 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177868 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12134 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164888 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81301 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87650 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169337 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167787 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33055 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133972 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17633 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P406169 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P406169 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P406169 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P406169 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 831 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16769 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35243 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34103 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97858 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67843 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63263 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35042 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31488 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10787990 - -
          Affymetrix Rat Gene 1.0 ST Array 10787992 - -
          Affymetrix Rat Gene 1.0 ST Array 10787996 - -
          Affymetrix Rat Gene 1.0 ST Array 10787991 - -
          Affymetrix Rat Gene 1.0 ST Array 10788001 - -
          Affymetrix Rat Gene 1.0 ST Array 10787989 - -
          Affymetrix Rat Gene 1.0 ST Array 10787998 - -
          Affymetrix Rat Gene 1.0 ST Array 10788000 - -
          Affymetrix Rat Gene 1.0 ST Array 10787999 - -
          Affymetrix Rat Gene 1.0 ST Array 10787995 - -
          Affymetrix Rat Gene 1.0 ST Array 10787993 - -
          Affymetrix Rat Gene 1.0 ST Array 10787997 - -
          Affymetrix Rat Gene 1.1 ST Array 10787988 - -
          Rat Genome 230 2.0 Array 1369431_at - -
          Rat Genome 230 2.0 Array 1369431_PM_at - -
          Affymetrix HT Rat Focus Array 1369431_at - -
          Affymetrix HT Rat Focus Array 1369431_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600004055.rn.1 TC1600000455.rn.1 RTA TC
          Rat Expression Set 230 1369431_at - -
          Rat Expression Set 230 1369431_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10787988 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22095780 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370216 - -
          RatRef-12 Expression BeadChip ILMN_67365 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline