Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576051_m1
          See other STX12 GE Assays ›
          Gene Symbol:
          Stx12
          Gene Name:
          syntaxin 12
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 151044050 - 151072888 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00576051_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stx12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022939.2 NP_075228.2 2 - 3 269 75
          XM_006239066.2 XP_006239128.1 2 - 3 313 75
          GenBank mRNA AF035632.1 - 2 - 3 185 75
          CB581551.1 - 2 - 3 258 75
          CK602227.1 - 2 - 3 269 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003763.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stx12

          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          vesicle fusion
          cholesterol efflux
          vesicle docking
          protein stabilization
          SNARE binding
          SNAP receptor activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P353685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253643 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111368 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138435 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40419 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137690 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69353 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32260 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P353685 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P353685 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P353685 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P251756 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P353685 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12343 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30843 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37398 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28990 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872645 - -
          Affymetrix Rat Gene 1.0 ST Array 10872650 - -
          Affymetrix Rat Gene 1.0 ST Array 10872647 - -
          Affymetrix Rat Gene 1.0 ST Array 10872649 - -
          Affymetrix Rat Gene 1.0 ST Array 10872653 - -
          Affymetrix Rat Gene 1.0 ST Array 10872651 - -
          Affymetrix Rat Gene 1.0 ST Array 10872646 - -
          Affymetrix Rat Gene 1.0 ST Array 10872643 - -
          Affymetrix Rat Gene 1.0 ST Array 10872648 - -
          Affymetrix Rat Gene 1.0 ST Array 10872652 - -
          Affymetrix Rat Gene 1.0 ST Array 10872644 - -
          Affymetrix Rat Gene 1.1 ST Array 10872642 - -
          Rat Genome 230 2.0 Array 1370192_PM_at - -
          Rat Genome 230 2.0 Array 1370192_at - -
          Affymetrix HT Rat Focus Array 1370192_at - -
          Affymetrix HT Rat Focus Array 1370192_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500028122.rn.1 TC0500003763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028124.rn.1 TC0500003763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028121.rn.1 TC0500003763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028116.rn.1 TC0500003763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028120.rn.1 TC0500003763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028123.rn.1 TC0500003763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028118.rn.1 TC0500003763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028119.rn.1 TC0500003763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028114.rn.1 TC0500003763.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028115.rn.1 TC0500003763.rn.1 RTA TC
          Rat Expression Set 230 1370192_at - -
          Rat Expression Set 230 1370192_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872642 - -
          Rat Genome U34 Set AF035632_s_at - -
          Rat Genome U34 Set AF044581_at - -
          Rat Genome U34 Set AF035632_s_at - -
          Rat Genome U34 Set AF044581_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22412446 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352795 - -
          RatRef-12 Expression BeadChip ILMN_56453 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline