Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576070_m1
          See other PDZD2 GE Assays ›
          Gene Symbol:
          Pdzd2
          Gene Name:
          PDZ domain containing 2
          Gene Aliases:
          Papin, Pdzk3
          Chromosome Location:
          Chr.2: 62399748 - 62520448 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00576070_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdzd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022940.1 NP_075229.1 8139 99
          GenBank mRNA AF169411.1 - 8139 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003093.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pdzd2

          Panther Classification:

          Molecular Function -

          interleukin superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          intracellular signal transduction
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P226928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105809 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41743 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52559 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152381 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21252 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123000 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136527 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83979 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109932 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160286 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176831 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80966 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7619 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P226928 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P226928 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P226928 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P226928 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701916234 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70000 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102534 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62398 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98557 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90341 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34463 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6529 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73024 - -
          Agilent Rat cDNA Microarray (G4105A) 701916234 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10822018 - -
          Affymetrix Rat Gene 1.0 ST Array 10822011 - -
          Affymetrix Rat Gene 1.0 ST Array 10822012 - -
          Affymetrix Rat Gene 1.0 ST Array 10822022 - -
          Affymetrix Rat Gene 1.0 ST Array 10822017 - -
          Affymetrix Rat Gene 1.0 ST Array 10822009 - -
          Affymetrix Rat Gene 1.0 ST Array 10822008 - -
          Affymetrix Rat Gene 1.0 ST Array 10822024 - -
          Affymetrix Rat Gene 1.0 ST Array 10822019 - -
          Affymetrix Rat Gene 1.0 ST Array 10822015 - -
          Affymetrix Rat Gene 1.0 ST Array 10822020 - -
          Affymetrix Rat Gene 1.0 ST Array 10822016 - -
          Affymetrix Rat Gene 1.0 ST Array 10822023 - -
          Affymetrix Rat Gene 1.0 ST Array 10822013 - -
          Affymetrix Rat Gene 1.0 ST Array 10822010 - -
          Affymetrix Rat Gene 1.1 ST Array 10822007 - -
          Rat Genome 230 2.0 Array 1369327_PM_at - -
          Rat Genome 230 2.0 Array 1369327_at - -
          Affymetrix HT Rat Focus Array 1369327_PM_at - -
          Affymetrix HT Rat Focus Array 1369327_at - -
          Affymetrix Rat Clariom S Assay PSR0200021660.rn.1 TC0200003093.rn.1 RTA TC
          Rat Expression Set 230 1369327_at - -
          Rat Expression Set 230 1369327_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10822007 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21266779 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351111 - -
          RatRef-12 Expression BeadChip ILMN_55054 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline