Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576087_m1
          See other BCHE GE Assays ›
          Gene Symbol:
          Bche
          Gene Name:
          butyrylcholinesterase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 171100137 - 171196186 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00576087_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bche
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022942.1 NP_075231.1 2 - 3 1589 93
          XM_008761112.1 XP_008759334.1 1 - 2 1666 93
          GenBank mRNA AF244349.1 - 2 - 3 1589 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003873.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bche

          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to nutrient
          learning
          negative regulation of cell proliferation
          neuroblast differentiation
          choline metabolic process
          response to drug
          response to alkaloid
          negative regulation of synaptic transmission
          response to glucocorticoid
          response to folic acid
          acetylcholinesterase activity
          cholinesterase activity
          choline binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683344 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2157 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68084 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113952 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42088 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16045 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104966 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139885 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95120 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12345 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12345 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12345 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700810345 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99504 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37411 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98755 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66422 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35162 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41767 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24283 - -
          Agilent Rat cDNA Microarray (G4105A) 700810345 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823682 - -
          Affymetrix Rat Gene 1.0 ST Array 10823680 - -
          Affymetrix Rat Gene 1.0 ST Array 10823681 - -
          Affymetrix Rat Gene 1.0 ST Array 10823679 - -
          Affymetrix Rat Gene 1.1 ST Array 10823678 - -
          Rat Genome 230 2.0 Array 1387832_at - -
          Rat Genome 230 2.0 Array 1387832_PM_at - -
          Affymetrix HT Rat Focus Array 1387832_PM_at - -
          Affymetrix HT Rat Focus Array 1387832_at - -
          Affymetrix Rat Clariom S Assay PSR0200025938.rn.1 TC0200003873.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200025939.rn.1 TC0200003873.rn.1 RTA TC
          Rat Expression Set 230 1387832_at - -
          Rat Expression Set 230 1387832_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10823678 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21585817 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349912 - -
          RatRef-12 Expression BeadChip ILMN_62643 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline