Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576163_m1
          See other CLPB GE Assays ›
          Gene Symbol:
          Clpb
          Gene Name:
          ClpB caseinolytic peptidase B homolog (E. coli)
          Gene Aliases:
          Skd3
          Chromosome Location:
          Chr.1: 166739372 - 166866095 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00576163_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clpb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022947.1 NP_075236.1 7 - 8 1033 103
          XM_008759796.1 XP_008758018.1 8 - 9 1387 103
          GenBank mRNA AB027570.1 - 7 - 8 1033 103
          BC070917.1 - 7 - 8 996 7105142 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100008047.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clpb

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          cellular response to heat
          molecular_function
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P539724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499663 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51667 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116435 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82211 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132767 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71821 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32513 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61871 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154123 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38037 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135200 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165009 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14100 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76966 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P539724 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P539724 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P539724 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P539724 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701233563 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88611 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39296 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28925 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23941 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14222 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78240 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60564 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93131 - -
          Agilent Rat cDNA Microarray (G4105A) 701233563 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10709244 - -
          Affymetrix Rat Gene 1.0 ST Array 10709249 - -
          Affymetrix Rat Gene 1.0 ST Array 10709246 - -
          Affymetrix Rat Gene 1.0 ST Array 10709245 - -
          Affymetrix Rat Gene 1.0 ST Array 10709250 - -
          Affymetrix Rat Gene 1.0 ST Array 10709247 - -
          Affymetrix Rat Gene 1.0 ST Array 10709238 - -
          Affymetrix Rat Gene 1.0 ST Array 10709248 - -
          Affymetrix Rat Gene 1.0 ST Array 10709252 - -
          Affymetrix Rat Gene 1.0 ST Array 10709240 - -
          Affymetrix Rat Gene 1.0 ST Array 10709243 - -
          Affymetrix Rat Gene 1.0 ST Array 10709254 - -
          Affymetrix Rat Gene 1.0 ST Array 10709251 - -
          Affymetrix Rat Gene 1.0 ST Array 10709242 - -
          Affymetrix Rat Gene 1.0 ST Array 10709253 - -
          Affymetrix Rat Gene 1.0 ST Array 10709239 - -
          Affymetrix Rat Gene 1.1 ST Array 10709237 - -
          Rat Genome 230 2.0 Array 1367997_at - -
          Rat Genome 230 2.0 Array 1367997_PM_at - -
          Affymetrix HT Rat Focus Array 1367997_PM_at - -
          Affymetrix HT Rat Focus Array 1367997_at - -
          Affymetrix Rat Clariom S Assay PSR0100065383.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065394.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065408.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065407.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065388.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065397.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065399.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065393.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065402.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065379.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065381.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065398.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065401.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065404.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065387.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065390.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065395.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065396.rn.1 TC0100008047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100065400.rn.1 TC0100008047.rn.1 RTA TC
          Rat Expression Set 230 1367997_PM_at - -
          Rat Expression Set 230 1367997_at - -
          Affymetrix Rat Gene 1.0 ST Array 10709237 - -
          Rat Genome U34 Set rc_AA891445_at - -
          Rat Genome U34 Set rc_AA875411_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22338263 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376393 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline