Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576196_m1
          See other PPP1R10 GE Assays ›
          Gene Symbol:
          Ppp1r10
          Gene Name:
          protein phosphatase 1, regulatory subunit 10
          Gene Aliases:
          Fb19, Pnuts
          Chromosome Location:
          Chr.20: 3329677 - 3344768 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00576196_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp1r10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022951.2 NP_075240.1 6 - 7 938 64
          XM_008772763.1 XP_008770985.1 5 - 6 1622 64
          XM_008772764.1 XP_008770986.1 6 - 7 1420 64
          GenBank mRNA AF040954.1 - 5 - 6 901 64
          BC134805.1 - 5 - 6 911 8362313 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001030.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp1r10

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          negative regulation of catalytic activity
          DNA binding
          RNA binding
          protein phosphatase inhibitor activity
          protein phosphatase 1 binding
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P497323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516651 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162536 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18252 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P497323 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P497323 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P497323 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P497323 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700513474 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702388328 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701419686 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700814394 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9242 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63475 - -
          Agilent Rat cDNA Microarray (G4105A) 700513474 - -
          Agilent Rat cDNA Microarray (G4105A) 700814394 - -
          Agilent Rat cDNA Microarray (G4105A) 701419686 - -
          Agilent Rat cDNA Microarray (G4105A) 702388328 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10830992 - -
          Affymetrix Rat Gene 1.0 ST Array 10831003 - -
          Affymetrix Rat Gene 1.0 ST Array 10830991 - -
          Affymetrix Rat Gene 1.0 ST Array 10831005 - -
          Affymetrix Rat Gene 1.0 ST Array 10831010 - -
          Affymetrix Rat Gene 1.0 ST Array 10831001 - -
          Affymetrix Rat Gene 1.0 ST Array 10831008 - -
          Affymetrix Rat Gene 1.0 ST Array 10830995 - -
          Affymetrix Rat Gene 1.0 ST Array 10830993 - -
          Affymetrix Rat Gene 1.0 ST Array 10831009 - -
          Affymetrix Rat Gene 1.0 ST Array 10830996 - -
          Affymetrix Rat Gene 1.0 ST Array 10830998 - -
          Affymetrix Rat Gene 1.0 ST Array 10830997 - -
          Affymetrix Rat Gene 1.0 ST Array 10831002 - -
          Affymetrix Rat Gene 1.0 ST Array 10831000 - -
          Affymetrix Rat Gene 1.0 ST Array 10831004 - -
          Affymetrix Rat Gene 1.0 ST Array 10831007 - -
          Affymetrix Rat Gene 1.0 ST Array 10830990 - -
          Affymetrix Rat Gene 1.0 ST Array 10830999 - -
          Affymetrix Rat Gene 1.0 ST Array 10830994 - -
          Affymetrix Rat Gene 1.0 ST Array 10831006 - -
          Affymetrix Rat Gene 1.1 ST Array 10830989 - -
          Rat Genome 230 2.0 Array 1386971_at - -
          Rat Genome 230 2.0 Array 1386971_PM_at - -
          Affymetrix HT Rat Focus Array 1386971_at - -
          Affymetrix HT Rat Focus Array 1386971_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000008291.rn.1 TC2000001030.rn.1 RTA TC
          Rat Expression Set 230 1386971_at - -
          Rat Expression Set 230 1386971_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10830989 - -
          Rat Genome U34 Set AF040954_at - -
          Rat Genome U34 Set rc_AI012268_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20877822 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355426 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline