Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576382_m1
          See other RHAG GE Assays ›
          Gene Symbol:
          Rhag
          Gene Name:
          Rh-associated glycoprotein
          Gene Aliases:
          Rh50
          Chromosome Location:
          Chr.9: 23465190 - 23493081 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00576382_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rhag
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023022.2 NP_075411.2 3 - 4 580 111
          GenBank mRNA AB015194.1 - 3 - 4 553 111
          AF531097.1 - 3 - 4 580 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001676.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rhag

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular ion homeostasis
          biological_process
          carbon dioxide transport
          ammonium transport
          erythrocyte development
          multicellular organismal iron ion homeostasis
          ammonium transmembrane transport
          molecular_function
          ammonium transmembrane transporter activity
          ankyrin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272167 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34503 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7741 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13882 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13882 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13882 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P271810 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13882 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10926809 - -
          Affymetrix Rat Gene 1.0 ST Array 10926818 - -
          Affymetrix Rat Gene 1.0 ST Array 10926775 - -
          Affymetrix Rat Gene 1.0 ST Array 10926814 - -
          Affymetrix Rat Gene 1.0 ST Array 10926771 - -
          Affymetrix Rat Gene 1.0 ST Array 10926781 - -
          Affymetrix Rat Gene 1.0 ST Array 10926812 - -
          Affymetrix Rat Gene 1.0 ST Array 10926815 - -
          Affymetrix Rat Gene 1.0 ST Array 10926813 - -
          Affymetrix Rat Gene 1.0 ST Array 10926772 - -
          Affymetrix Rat Gene 1.0 ST Array 10926770 - -
          Affymetrix Rat Gene 1.0 ST Array 10926811 - -
          Affymetrix Rat Gene 1.0 ST Array 10926810 - -
          Affymetrix Rat Gene 1.0 ST Array 10926777 - -
          Affymetrix Rat Gene 1.0 ST Array 10926779 - -
          Affymetrix Rat Gene 1.0 ST Array 10926774 - -
          Affymetrix Rat Gene 1.0 ST Array 10926773 - -
          Affymetrix Rat Gene 1.0 ST Array 10926807 - -
          Affymetrix Rat Gene 1.0 ST Array 10926776 - -
          Affymetrix Rat Gene 1.0 ST Array 10926816 - -
          Affymetrix Rat Gene 1.0 ST Array 10926808 - -
          Affymetrix Rat Gene 1.0 ST Array 10926778 - -
          Affymetrix Rat Gene 1.1 ST Array 10926769 - -
          Affymetrix Rat Gene 1.1 ST Array 10926806 - -
          Rat Genome 230 2.0 Array 1387522_at - -
          Rat Genome 230 2.0 Array 1387522_PM_at - -
          Affymetrix HT Rat Focus Array 1387522_PM_at - -
          Affymetrix HT Rat Focus Array 1387522_at - -
          Affymetrix Rat Clariom S Assay PSR0900013759.rn.1 TC0900001676.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013761.rn.1 TC0900001676.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013758.rn.1 TC0900001676.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013760.rn.1 TC0900001676.rn.1 RTA TC
          Rat Expression Set 230 1387522_PM_at - -
          Rat Expression Set 230 1387522_at - -
          Affymetrix Rat Gene 1.0 ST Array 10926769 - -
          Affymetrix Rat Gene 1.0 ST Array 10926806 - -
          Rat Genome U34 Set AB015194_at - -
          Rat Genome U34 Set rc_AA963110_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21135652 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361605 - -
          RatRef-12 Expression BeadChip ILMN_53164 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline