Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576558_m1
          See other KCNK10 GE Assays ›
          Gene Symbol:
          Kcnk10
          Gene Name:
          potassium channel, two pore domain subfamily K, member 10
          Gene Aliases:
          TREK2
          Chromosome Location:
          Chr.6: 122417079 - 122547485 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00576558_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnk10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023096.2 NP_075584.1 2 - 3 756 119
          XM_008764847.1 XP_008763069.1 2 - 3 1476 119
          GenBank mRNA AF196965.1 - 2 - 3 567 119
          AF385401.1 - 2 - 3 756 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002996.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnk10

          Panther Classification:

          Molecular Function -

          ion channel transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of ion transmembrane transport
          potassium ion transmembrane transport
          voltage-gated ion channel activity
          potassium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P109118 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P363291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906585 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126542 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70699 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20283 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44195 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109294 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162630 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162575 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130613 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167596 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52482 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117442 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P363291 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109118 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109118 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P363291 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P109118 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P363291 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P363291 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57275 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81098 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5263 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86165 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45140 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7039 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86405 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67570 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31878 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48685 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10891527 - -
          Affymetrix Rat Gene 1.0 ST Array 10891525 - -
          Affymetrix Rat Gene 1.0 ST Array 10891526 - -
          Affymetrix Rat Gene 1.0 ST Array 10891529 - -
          Affymetrix Rat Gene 1.0 ST Array 10891524 - -
          Affymetrix Rat Gene 1.0 ST Array 10891522 - -
          Affymetrix Rat Gene 1.0 ST Array 10891523 - -
          Affymetrix Rat Gene 1.0 ST Array 10891528 - -
          Affymetrix Rat Gene 1.0 ST Array 10891521 - -
          Affymetrix Rat Gene 1.1 ST Array 10891520 - -
          Rat Genome 230 2.0 Array 1369232_at - -
          Rat Genome 230 2.0 Array 1369232_PM_at - -
          Rat Genome 230 2.0 Array 1369233_at - -
          Rat Genome 230 2.0 Array 1369233_PM_at - -
          Affymetrix HT Rat Focus Array 1369232_PM_at - -
          Affymetrix HT Rat Focus Array 1369233_PM_at - -
          Affymetrix HT Rat Focus Array 1369232_at - -
          Affymetrix HT Rat Focus Array 1369233_at - -
          Affymetrix Rat Clariom S Assay PSR0600021073.rn.1 TC0600002996.rn.1 RTA TC
          Rat Expression Set 230 1369233_PM_at - -
          Rat Expression Set 230 1369233_at - -
          Rat Expression Set 230 1369232_PM_at - -
          Rat Expression Set 230 1369232_at - -
          Affymetrix Rat Gene 1.0 ST Array 10891520 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21380544 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039953 - -
          RatRef-12 Expression BeadChip ILMN_55247 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline