Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576578_m1
          See other VTI1A GE Assays ›
          Gene Symbol:
          Vti1a
          Gene Name:
          vesicle transport through interaction with t-SNAREs 1A
          Gene Aliases:
          Vti1
          Chromosome Location:
          Chr.1: 276310072 - 276508635 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00576578_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vti1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023101.1 NP_075589.1 4 - 5 373 62
          GenBank mRNA AF262221.1 - 4 - 5 373 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003845.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vti1a

          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to vacuole
          intracellular protein transport
          ER to Golgi vesicle-mediated transport
          intra-Golgi vesicle-mediated transport
          Golgi to vacuole transport
          vesicle-mediated transport
          retrograde transport, endosome to Golgi
          vesicle fusion with Golgi apparatus
          voluntary musculoskeletal movement
          membrane fusion
          Golgi ribbon formation
          SNARE binding
          SNAP receptor activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835257 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43544 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125538 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110086 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50005 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61822 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171086 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80597 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130629 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163658 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114760 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16524 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110060 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118000 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144310 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106440 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26464 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89931 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83975 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69541 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80581 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52265 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7593 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44699 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29472 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12365 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12365 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12365 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12365 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701621014 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68659 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37427 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11277 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37281 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3175 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67497 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51205 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43513 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37020 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31592 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68331 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25391 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48541 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55564 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63204 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74778 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48391 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47675 - -
          Agilent Rat cDNA Microarray (G4105A) 701621014 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940531 - -
          Affymetrix Rat Gene 1.1 ST Array 10940531 - -
          Rat Genome 230 2.0 Array 1368889_at - -
          Rat Genome 230 2.0 Array 1379980_at - -
          Rat Genome 230 2.0 Array 1368889_PM_at - -
          Rat Genome 230 2.0 Array 1379980_PM_at - -
          Affymetrix HT Rat Focus Array 1368889_at - -
          Affymetrix HT Rat Focus Array 1368889_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100031427.rn.1 TC0100003845.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100031432.rn.1 TC0100003845.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100031433.rn.1 TC0100003845.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100031430.rn.1 TC0100003845.rn.1 RTA TC
          Rat Expression Set 230 1368889_at - -
          Rat Expression Set 230 1368889_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940531 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20708653 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376721 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline