Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576694_m1
          See other CDX2 GE Assays ›
          Gene Symbol:
          Cdx2
          Gene Name:
          caudal type homeo box 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 9464026 - 9470565 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00576694_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdx2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023963.1 NP_076453.1 1 - 2 651 89
          XM_008768999.1 XP_008767221.1 1 - 2 542 89
          GenBank mRNA AJ278466.1 - 1 - 2 651 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000159.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdx2

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          blood vessel development
          blastocyst development
          trophectodermal cell differentiation
          placenta development
          regulation of transcription from RNA polymerase II promoter
          pattern specification process
          positive regulation of cell proliferation
          endosome to lysosome transport
          anterior/posterior pattern specification
          cell differentiation
          somatic stem cell population maintenance
          establishment or maintenance of epithelial cell apical/basal polarity
          positive regulation of cell differentiation
          positive regulation of transcription, DNA-templated
          labyrinthine layer development
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564945 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128174 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157803 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157803 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157803 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P655689 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22027 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756491 - -
          Affymetrix Rat Gene 1.0 ST Array 10756488 - -
          Affymetrix Rat Gene 1.0 ST Array 10756490 - -
          Affymetrix Rat Gene 1.0 ST Array 10756489 - -
          Affymetrix Rat Gene 1.1 ST Array 10756487 - -
          Rat Genome 230 2.0 Array 1387726_PM_at - -
          Rat Genome 230 2.0 Array 1387726_at - -
          Affymetrix HT Rat Focus Array 1387726_at - -
          Affymetrix HT Rat Focus Array 1387726_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200001131.rn.1 TC1200000159.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200001129.rn.1 TC1200000159.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200001130.rn.1 TC1200000159.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200001136.rn.1 TC1200000159.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200001135.rn.1 TC1200000159.rn.1 RTA TC
          Rat Expression Set 230 1387726_at - -
          Rat Expression Set 230 1387726_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10756487 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20863565 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368698 - -
          RatRef-12 Expression BeadChip ILMN_66195 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline