Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577115_m1
          See other LIMK2 GE Assays ›
          Gene Symbol:
          Limk2
          Gene Name:
          LIM domain kinase 2
          Gene Aliases:
          Limk2b, Link2
          Chromosome Location:
          Chr.14: 83564048 - 83641995 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577115_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Limk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024135.2 NP_077049.2 7 - 8 922 60
          XM_006251236.2 XP_006251298.1 6 - 7 2239 60
          XM_008770275.1 XP_008768497.1 7 - 8 1025 60
          XM_008770276.1 XP_008768498.1 6 - 7 2056 60
          XM_008770277.1 XP_008768499.1 6 - 7 2239 60
          XM_008770278.1 XP_008768500.1 7 - 8 1024 60
          XM_008770279.1 XP_008768501.1 7 - 8 1024 60
          GenBank mRNA BC065578.1 - 6 - 7 1107 5623751 60
          D31874.1 - 7 - 8 922 60
          D31875.1 - 6 - 7 1151 60
          D31876.1 - 6 - 7 1151 60
          D31877.1 - 6 - 7 1275 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002125.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Limk2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          spermatogenesis
          spermatid development
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          zinc ion binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P461451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620229 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126235 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178589 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66893 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91837 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68618 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42285 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143851 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P461451 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P461451 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P461451 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P461451 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P593993 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23518 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100135 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7803 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82807 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10777942 - -
          Affymetrix Rat Gene 1.0 ST Array 10777949 - -
          Affymetrix Rat Gene 1.0 ST Array 10777945 - -
          Affymetrix Rat Gene 1.0 ST Array 10777935 - -
          Affymetrix Rat Gene 1.0 ST Array 10777948 - -
          Affymetrix Rat Gene 1.0 ST Array 10777937 - -
          Affymetrix Rat Gene 1.0 ST Array 10777943 - -
          Affymetrix Rat Gene 1.0 ST Array 10777940 - -
          Affymetrix Rat Gene 1.0 ST Array 10777946 - -
          Affymetrix Rat Gene 1.0 ST Array 10777938 - -
          Affymetrix Rat Gene 1.0 ST Array 10777941 - -
          Affymetrix Rat Gene 1.0 ST Array 10777933 - -
          Affymetrix Rat Gene 1.0 ST Array 10777932 - -
          Affymetrix Rat Gene 1.0 ST Array 10777944 - -
          Affymetrix Rat Gene 1.0 ST Array 10777934 - -
          Affymetrix Rat Gene 1.0 ST Array 10777939 - -
          Affymetrix Rat Gene 1.0 ST Array 10777929 - -
          Affymetrix Rat Gene 1.0 ST Array 10777936 - -
          Affymetrix Rat Gene 1.0 ST Array 10777947 - -
          Affymetrix Rat Gene 1.0 ST Array 10777930 - -
          Affymetrix Rat Gene 1.0 ST Array 10777931 - -
          Affymetrix Rat Gene 1.1 ST Array 10777928 - -
          Rat Genome 230 2.0 Array 1387090_a_at - -
          Rat Genome 230 2.0 Array 1387090_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387090_a_at - -
          Affymetrix HT Rat Focus Array 1387090_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1400015584.rn.1 TC1400002125.rn.1 RTA TC
          Rat Expression Set 230 1387090_a_at - -
          Rat Expression Set 230 1387090_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10777928 - -
          Rat Genome U34 Set D31874_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20827097 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355049 - -
          RatRef-12 Expression BeadChip ILMN_65547 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0