Search
Search
| Gene Symbol: Arf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024152.2 | NP_077066.1 | 1 | 567 | 72 | |
| GenBank mRNA | BC091146.1 | - | 1 | 567 | 7309902 | 72 |
| L12385.1 | - | 1 | 556 | 72 | ||
Set Membership: |
|