Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577450_m1
          See other GCH1 GE Assays ›
          Gene Symbol:
          Gch1
          Gene Name:
          GTP cyclohydrolase 1
          Gene Aliases:
          Gch
          Chromosome Location:
          Chr.15: 23935011 - 23968971 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577450_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gch1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024356.1 NP_077332.1 3 - 4 605 108
          GenBank mRNA FQ220953.1 - 3 - 4 592 108
          M58364.1 - 3 - 4 605 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001485.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gch1

          Panther Classification:

          Molecular Function -

          hydrolase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          tetrahydrobiopterin biosynthetic process
          metabolic process
          regulation of blood pressure
          regulation of lung blood pressure
          response to lipopolysaccharide
          response to interferon-gamma
          response to tumor necrosis factor
          7,8-dihydroneopterin 3'-triphosphate biosynthetic process
          vasodilation
          dopamine biosynthetic process
          pteridine-containing compound biosynthetic process
          negative regulation of blood pressure
          tetrahydrofolate biosynthetic process
          response to pain
          neuromuscular process controlling posture
          positive regulation of nitric-oxide synthase activity
          dihydrobiopterin metabolic process
          protein homooligomerization
          protein heterooligomerization
          GTP cyclohydrolase I activity
          calcium ion binding
          protein binding
          GTP binding
          zinc ion binding
          GTP-dependent protein binding
          protein homodimerization activity
          coenzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P138216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655198 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12944 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108202 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112870 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9835 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138216 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138216 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P138216 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P390495 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701878715 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94339 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17292 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4344 - -
          Agilent Rat cDNA Microarray (G4105A) 701878715 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10782923 - -
          Affymetrix Rat Gene 1.0 ST Array 10782922 - -
          Affymetrix Rat Gene 1.0 ST Array 10782926 - -
          Affymetrix Rat Gene 1.0 ST Array 10782925 - -
          Affymetrix Rat Gene 1.0 ST Array 10782920 - -
          Affymetrix Rat Gene 1.0 ST Array 10782921 - -
          Affymetrix Rat Gene 1.0 ST Array 10782924 - -
          Affymetrix Rat Gene 1.1 ST Array 10782919 - -
          Rat Genome 230 2.0 Array 1373513_PM_at - -
          Rat Genome 230 2.0 Array 1368503_at - -
          Rat Genome 230 2.0 Array 1368503_PM_at - -
          Rat Genome 230 2.0 Array 1387221_PM_at - -
          Rat Genome 230 2.0 Array 1387221_at - -
          Rat Genome 230 2.0 Array 1373513_at - -
          Affymetrix HT Rat Focus Array 1387221_PM_at - -
          Affymetrix HT Rat Focus Array 1368503_at - -
          Affymetrix HT Rat Focus Array 1387221_at - -
          Affymetrix HT Rat Focus Array 1368503_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500009410.rn.1 TC1500001485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500009412.rn.1 TC1500001485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500009411.rn.1 TC1500001485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500009419.rn.1 TC1500001485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500009420.rn.1 TC1500001485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500009415.rn.1 TC1500001485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500009409.rn.1 TC1500001485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500009418.rn.1 TC1500001485.rn.1 RTA TC
          Rat Expression Set 230 1387221_at - -
          Rat Expression Set 230 1387221_PM_at - -
          Rat Expression Set 230 1368503_PM_at - -
          Rat Expression Set 230 1368503_at - -
          Affymetrix Rat Gene 1.0 ST Array 10782919 - -
          Rat Genome U34 Set M58364_at - -
          Rat Genome U34 Set rc_AA943102_s_at - -
          Rat Genome U34 Set rc_AI043643_at - -
          Rat Genome U34 Set E03424cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21398215 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372022 - -
          RatRef-12 Expression BeadChip ILMN_65394 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0