Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577462_m1
          See other HTT GE Assays ›
          Gene Symbol:
          Htt
          Gene Name:
          huntingtin
          Gene Aliases:
          Hd, Hdh
          Chromosome Location:
          Chr.14: 81105139 - 81254637 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577462_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Htt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024357.3 NP_077333.2 19 - 20 2741 74
          XM_006251230.2 XP_006251292.1 18 - 19 2679 74
          XM_006251232.2 XP_006251294.1 19 - 20 2727 74
          XM_008770274.1 XP_008768496.1 17 - 18 2193 74
          GenBank mRNA U18650.1 - 19 - 20 2585 74

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the U18650.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Htt

          Gene Ontology Categories:

          Function(s) Process(es)

          urea cycle
          citrulline metabolic process
          establishment of mitotic spindle orientation
          protein import into nucleus
          mitochondrial transport
          ER to Golgi vesicle-mediated transport
          retrograde vesicle-mediated transport, Golgi to ER
          apoptotic process
          mitochondrion organization
          endoplasmic reticulum organization
          Golgi organization
          dopamine receptor signaling pathway
          spermatogenesis
          gastrulation
          central nervous system development
          brain development
          cell aging
          learning or memory
          learning
          grooming behavior
          locomotory behavior
          axo-dendritic transport
          associative learning
          determination of adult lifespan
          visual learning
          anatomical structure morphogenesis
          anterior/posterior pattern specification
          endosomal transport
          lactate biosynthetic process from pyruvate
          quinolinate biosynthetic process
          striatum development
          olfactory lobe development
          neural plate formation
          neurogenesis
          peptide hormone secretion
          insulin secretion
          positive regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
          regulation of protein phosphatase type 2A activity
          social behavior
          hormone metabolic process
          negative regulation of apoptotic process
          negative regulation of neuron apoptotic process
          regulation of mitochondrial membrane permeability
          vesicle transport along microtubule
          regulation of synaptic plasticity
          paraxial mesoderm formation
          animal organ development
          neuron development
          mRNA transport
          neuron apoptotic process
          response to calcium ion
          regulation of mitochondrial membrane potential
          L-glutamate import
          iron ion homeostasis
          negative regulation of neuron death
          negative regulation of cysteine-type endopeptidase activity
          negative regulation of extrinsic apoptotic signaling pathway
          p53 binding
          receptor binding
          protein binding
          transcription factor binding
          dynactin binding
          identical protein binding
          ion channel binding
          dynein intermediate chain binding
          beta-tubulin binding
          diazepam binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P316136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588245 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77291 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100397 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103619 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17449 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152294 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88723 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15284 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39118 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62812 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72341 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180286 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61305 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169413 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108539 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43779 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P316136 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P316136 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P316136 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P316136 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P161033 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701606012 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15750 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63387 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36189 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36422 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91264 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86123 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100997 - -
          Agilent Rat cDNA Microarray (G4105A) 701606012 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10777574 - -
          Affymetrix Rat Gene 1.0 ST Array 10777582 - -
          Affymetrix Rat Gene 1.0 ST Array 10777614 - -
          Affymetrix Rat Gene 1.0 ST Array 10777616 - -
          Affymetrix Rat Gene 1.0 ST Array 10777592 - -
          Affymetrix Rat Gene 1.0 ST Array 10777600 - -
          Affymetrix Rat Gene 1.0 ST Array 10777629 - -
          Affymetrix Rat Gene 1.0 ST Array 10777619 - -
          Affymetrix Rat Gene 1.0 ST Array 10777610 - -
          Affymetrix Rat Gene 1.0 ST Array 10777599 - -
          Affymetrix Rat Gene 1.0 ST Array 10777601 - -
          Affymetrix Rat Gene 1.0 ST Array 10777624 - -
          Affymetrix Rat Gene 1.0 ST Array 10777596 - -
          Affymetrix Rat Gene 1.0 ST Array 10777578 - -
          Affymetrix Rat Gene 1.0 ST Array 10777577 - -
          Affymetrix Rat Gene 1.0 ST Array 10777586 - -
          Affymetrix Rat Gene 1.0 ST Array 10777588 - -
          Affymetrix Rat Gene 1.0 ST Array 10777638 - -
          Affymetrix Rat Gene 1.0 ST Array 10777591 - -
          Affymetrix Rat Gene 1.0 ST Array 10777575 - -
          Affymetrix Rat Gene 1.0 ST Array 10777573 - -
          Affymetrix Rat Gene 1.0 ST Array 10777632 - -
          Affymetrix Rat Gene 1.0 ST Array 10777576 - -
          Affymetrix Rat Gene 1.0 ST Array 10777572 - -
          Affymetrix Rat Gene 1.0 ST Array 10777593 - -
          Affymetrix Rat Gene 1.0 ST Array 10777633 - -
          Affymetrix Rat Gene 1.0 ST Array 10777613 - -
          Affymetrix Rat Gene 1.0 ST Array 10777581 - -
          Affymetrix Rat Gene 1.0 ST Array 10777611 - -
          Affymetrix Rat Gene 1.0 ST Array 10777580 - -
          Affymetrix Rat Gene 1.0 ST Array 10777617 - -
          Affymetrix Rat Gene 1.0 ST Array 10777608 - -
          Affymetrix Rat Gene 1.0 ST Array 10777603 - -
          Affymetrix Rat Gene 1.0 ST Array 10777605 - -
          Affymetrix Rat Gene 1.0 ST Array 10777623 - -
          Affymetrix Rat Gene 1.0 ST Array 10777625 - -
          Affymetrix Rat Gene 1.0 ST Array 10777585 - -
          Affymetrix Rat Gene 1.0 ST Array 10777637 - -
          Affymetrix Rat Gene 1.0 ST Array 10777615 - -
          Affymetrix Rat Gene 1.0 ST Array 10777607 - -
          Affymetrix Rat Gene 1.0 ST Array 10777628 - -
          Affymetrix Rat Gene 1.0 ST Array 10777597 - -
          Affymetrix Rat Gene 1.0 ST Array 10777622 - -
          Affymetrix Rat Gene 1.0 ST Array 10777618 - -
          Affymetrix Rat Gene 1.0 ST Array 10777630 - -
          Affymetrix Rat Gene 1.0 ST Array 10777631 - -
          Affymetrix Rat Gene 1.0 ST Array 10777626 - -
          Affymetrix Rat Gene 1.0 ST Array 10777595 - -
          Affymetrix Rat Gene 1.0 ST Array 10777589 - -
          Affymetrix Rat Gene 1.0 ST Array 10777594 - -
          Affymetrix Rat Gene 1.0 ST Array 10777583 - -
          Affymetrix Rat Gene 1.0 ST Array 10777587 - -
          Affymetrix Rat Gene 1.0 ST Array 10777627 - -
          Affymetrix Rat Gene 1.0 ST Array 10777621 - -
          Affymetrix Rat Gene 1.0 ST Array 10777579 - -
          Affymetrix Rat Gene 1.0 ST Array 10777634 - -
          Affymetrix Rat Gene 1.0 ST Array 10777636 - -
          Affymetrix Rat Gene 1.0 ST Array 10777612 - -
          Affymetrix Rat Gene 1.0 ST Array 10777620 - -
          Affymetrix Rat Gene 1.0 ST Array 10777598 - -
          Affymetrix Rat Gene 1.0 ST Array 10777590 - -
          Affymetrix Rat Gene 1.0 ST Array 10777609 - -
          Affymetrix Rat Gene 1.0 ST Array 10777635 - -
          Affymetrix Rat Gene 1.0 ST Array 10777602 - -
          Affymetrix Rat Gene 1.0 ST Array 10777604 - -
          Affymetrix Rat Gene 1.0 ST Array 10777584 - -
          Affymetrix Rat Gene 1.0 ST Array 10777606 - -
          Affymetrix Rat Gene 1.1 ST Array 10777571 - -
          Rat Genome 230 2.0 Array 1395570_PM_at - -
          Rat Genome 230 2.0 Array 1370395_PM_at - -
          Rat Genome 230 2.0 Array 1395570_at - -
          Rat Genome 230 2.0 Array 1370395_at - -
          Affymetrix HT Rat Focus Array 1370395_at - -
          Affymetrix HT Rat Focus Array 1370395_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400014902.rn.1 TC1400002083.rn.1 RTA TC
          Rat Expression Set 230 1370395_at - -
          Rat Expression Set 230 1370395_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10777571 - -
          Rat Genome U34 Set U18650_at - -
          Rat Genome U34 Set U01022_s_at - -
          Rat Genome U34 Set U01022_s_at - -
          Rat Genome U34 Set U18650_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22363901 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360694 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline