Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577605_m1
          See other HR GE Assays ›
          Gene Symbol:
          Hr
          Gene Name:
          hair growth associated
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 52241801 - 52261280 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577605_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024364.3 NP_077340.3 18 - 19 4043 73
          XM_006252283.1 XP_006252345.1 15 - 16 3270 73
          XM_008770833.1 XP_008769055.1 15 - 16 3270 73
          GenBank mRNA U71293.1 - 15 - 16 3659 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000778.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hr

          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of transcription, DNA-templated
          protein heterooligomerization
          oxidation-reduction process
          negative regulation of nucleic acid-templated transcription
          DNA binding
          transcription corepressor activity
          oxidoreductase activity
          vitamin D receptor binding
          histone deacetylase binding
          metal ion binding
          thyroid hormone receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P321176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266420 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97750 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86500 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60858 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P321176 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P321176 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P321176 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P321176 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700493788 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17125 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78984 - -
          Agilent Rat cDNA Microarray (G4105A) 700493788 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10781395 - -
          Affymetrix Rat Gene 1.0 ST Array 10781384 - -
          Affymetrix Rat Gene 1.0 ST Array 10781396 - -
          Affymetrix Rat Gene 1.0 ST Array 10781392 - -
          Affymetrix Rat Gene 1.0 ST Array 10781390 - -
          Affymetrix Rat Gene 1.0 ST Array 10781379 - -
          Affymetrix Rat Gene 1.0 ST Array 10781388 - -
          Affymetrix Rat Gene 1.0 ST Array 10781394 - -
          Affymetrix Rat Gene 1.0 ST Array 10781380 - -
          Affymetrix Rat Gene 1.0 ST Array 10781382 - -
          Affymetrix Rat Gene 1.0 ST Array 10781397 - -
          Affymetrix Rat Gene 1.0 ST Array 10781383 - -
          Affymetrix Rat Gene 1.0 ST Array 10781391 - -
          Affymetrix Rat Gene 1.0 ST Array 10781386 - -
          Affymetrix Rat Gene 1.0 ST Array 10781387 - -
          Affymetrix Rat Gene 1.0 ST Array 10781385 - -
          Affymetrix Rat Gene 1.0 ST Array 10781381 - -
          Affymetrix Rat Gene 1.0 ST Array 10781393 - -
          Affymetrix Rat Gene 1.0 ST Array 10781389 - -
          Affymetrix Rat Gene 1.1 ST Array 10781378 - -
          Rat Genome 230 2.0 Array 1369336_PM_at - -
          Rat Genome 230 2.0 Array 1369336_at - -
          Affymetrix HT Rat Focus Array 1369336_at - -
          Affymetrix HT Rat Focus Array 1369336_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500005446.rn.1 TC1500000778.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005447.rn.1 TC1500000778.rn.1 RTA TC
          Rat Expression Set 230 1369336_PM_at - -
          Rat Expression Set 230 1369336_at - -
          Affymetrix Rat Gene 1.0 ST Array 10781378 - -
          Rat Genome U34 Set U71293_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22085078 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370396 - -
          RatRef-12 Expression BeadChip ILMN_69308 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline