Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577652_m1
          See other SLC6A1 GE Assays ›
          Gene Symbol:
          Slc6a1
          Gene Name:
          solute carrier family 6 (neurotransmitter transporter), member 1
          Gene Aliases:
          Gabt1, Gat1
          Chromosome Location:
          Chr.4: 146258834 - 146292176 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577652_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc6a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024371.2 NP_077347.1 5 - 6 732 94
          XM_006237059.1 XP_006237121.1 6 - 7 941 94
          XM_006237060.2 XP_006237122.1 6 - 7 3161 94
          GenBank mRNA M59742.1 - 5 - 6 732 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001592.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc6a1

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter transport
          learning
          response to toxic substance
          response to sucrose
          response to organic substance
          response to organonitrogen compound
          response to lead ion
          positive regulation of gamma-aminobutyric acid secretion
          response to organic cyclic compound
          response to purine-containing compound
          negative regulation of synaptic transmission, GABAergic
          response to estradiol
          response to cocaine
          protein homooligomerization
          response to calcium ion
          gamma-aminobutyric acid import
          transmembrane transport
          neurotransmitter:sodium symporter activity
          gamma-aminobutyric acid:sodium symporter activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P113691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549288 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159014 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P113691 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P113691 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P113691 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P113691 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701903954 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58547 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5408 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15600 - -
          Agilent Rat cDNA Microarray (G4105A) 701903954 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10857943 - -
          Affymetrix Rat Gene 1.0 ST Array 10857945 - -
          Affymetrix Rat Gene 1.0 ST Array 10857946 - -
          Affymetrix Rat Gene 1.0 ST Array 10857942 - -
          Affymetrix Rat Gene 1.0 ST Array 10857936 - -
          Affymetrix Rat Gene 1.0 ST Array 10857937 - -
          Affymetrix Rat Gene 1.0 ST Array 10857934 - -
          Affymetrix Rat Gene 1.0 ST Array 10857932 - -
          Affymetrix Rat Gene 1.0 ST Array 10857940 - -
          Affymetrix Rat Gene 1.0 ST Array 10857944 - -
          Affymetrix Rat Gene 1.0 ST Array 10857939 - -
          Affymetrix Rat Gene 1.0 ST Array 10857933 - -
          Affymetrix Rat Gene 1.0 ST Array 10857938 - -
          Affymetrix Rat Gene 1.0 ST Array 10857935 - -
          Affymetrix Rat Gene 1.0 ST Array 10857941 - -
          Affymetrix Rat Gene 1.1 ST Array 10857931 - -
          Rat Genome 230 2.0 Array 1368170_PM_at - -
          Rat Genome 230 2.0 Array 1368170_at - -
          Affymetrix HT Rat Focus Array 1368170_at - -
          Affymetrix HT Rat Focus Array 1368170_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400011341.rn.1 TC0400001592.rn.1 RTA TC
          Rat Expression Set 230 1368170_at - -
          Rat Expression Set 230 1368170_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10857931 - -
          Rat Genome U34 Set rc_AI228669_at - -
          Rat Genome U34 Set rc_AI228669_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21936475 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363429 - -
          RatRef-12 Expression BeadChip ILMN_59754 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline