Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577670_m1
          See other GALNT1 GE Assays ›
          Gene Symbol:
          Galnt1
          Gene Name:
          polypeptide N-acetylgalactosaminyltransferase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 16141064 - 16187271 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577670_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Galnt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024373.1 NP_077349.1 10 - 11 1639 67
          XM_006254471.2 XP_006254533.1 11 - 12 3060 67
          GenBank mRNA U35890.1 - 10 - 11 1639 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000154.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Galnt1

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein O-linked glycosylation
          protein O-linked glycosylation via serine
          protein O-linked glycosylation via threonine
          polypeptide N-acetylgalactosaminyltransferase activity
          manganese ion binding
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P524981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634666 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12874 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20435 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108697 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P524981 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P524981 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P524981 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P590280 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P524981 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700254034 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42369 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78631 - -
          Agilent Rat cDNA Microarray (G4105A) 700254034 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10800499 - -
          Affymetrix Rat Gene 1.0 ST Array 10800503 - -
          Affymetrix Rat Gene 1.0 ST Array 10800509 - -
          Affymetrix Rat Gene 1.0 ST Array 10800504 - -
          Affymetrix Rat Gene 1.0 ST Array 10800506 - -
          Affymetrix Rat Gene 1.0 ST Array 10800505 - -
          Affymetrix Rat Gene 1.0 ST Array 10800507 - -
          Affymetrix Rat Gene 1.0 ST Array 10800500 - -
          Affymetrix Rat Gene 1.0 ST Array 10800498 - -
          Affymetrix Rat Gene 1.0 ST Array 10800501 - -
          Affymetrix Rat Gene 1.0 ST Array 10800508 - -
          Affymetrix Rat Gene 1.0 ST Array 10800502 - -
          Affymetrix Rat Gene 1.1 ST Array 10800497 - -
          Rat Genome 230 2.0 Array 1399045_PM_at - -
          Rat Genome 230 2.0 Array 1381990_at - -
          Rat Genome 230 2.0 Array 1396345_at - -
          Rat Genome 230 2.0 Array 1383374_PM_at - -
          Rat Genome 230 2.0 Array 1369269_PM_at - -
          Rat Genome 230 2.0 Array 1369269_at - -
          Rat Genome 230 2.0 Array 1396345_PM_at - -
          Rat Genome 230 2.0 Array 1381990_PM_at - -
          Rat Genome 230 2.0 Array 1383374_at - -
          Rat Genome 230 2.0 Array 1399045_at - -
          Affymetrix HT Rat Focus Array 1369269_PM_at - -
          Affymetrix HT Rat Focus Array 1383374_PM_at - -
          Affymetrix HT Rat Focus Array 1383374_at - -
          Affymetrix HT Rat Focus Array 1396345_at - -
          Affymetrix HT Rat Focus Array 1399045_at - -
          Affymetrix HT Rat Focus Array 1369269_at - -
          Affymetrix HT Rat Focus Array 1399045_PM_at - -
          Affymetrix HT Rat Focus Array 1396345_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800001032.rn.1 TC1800000154.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800001031.rn.1 TC1800000154.rn.1 RTA TC
          Rat Expression Set 230 1399045_at - -
          Rat Expression Set 230 1369269_at - -
          Rat Expression Set 230 1399045_PM_at - -
          Rat Expression Set 230 1369269_PM_at - -
          Rat Expression Set 230 1383374_at - -
          Rat Expression Set 230 1383374_PM_at - -
          Rat Expression Set 230 1381990_PM_at - -
          Rat Expression Set 230 1396345_at - -
          Rat Expression Set 230 1381990_at - -
          Rat Expression Set 230 1396345_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10800497 - -
          Rat Genome U34 Set U35890_at - -
          Rat Genome U34 Set rc_AI043831_at - -
          Rat Genome U34 Set rc_AI045810_at - -
          Rat Genome U34 Set rc_AI045597_at - -
          Rat Genome U34 Set rc_AI236745_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21942790 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline