Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577740_m1
          See other GK GE Assays ›
          Gene Symbol:
          Gk
          Gene Name:
          glycerol kinase
          Gene Aliases:
          ASTP, Gyk
          Chromosome Location:
          Chr.X: 54227291 - 54303859 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577740_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024381.2 NP_077357.2 6 - 7 651 94
          XM_006256987.2 XP_006257049.1 6 - 7 685 94
          XM_006256988.2 XP_006257050.1 6 - 7 685 94
          XM_008773274.1 XP_008771496.1 7 - 8 750 94
          XM_008773275.1 XP_008771497.1 7 - 8 750 94
          XM_008773276.1 XP_008771498.1 7 - 8 750 94
          XM_008773278.1 XP_008771500.1 7 - 8 749 94
          GenBank mRNA D16102.1 - 6 - 7 654 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001797.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gk

          Panther Classification:

          Molecular Function -

          carbohydrate kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycerol metabolic process
          glycerol-3-phosphate metabolic process
          triglyceride metabolic process
          response to cold
          response to organic substance
          phosphorylation
          glycerol catabolic process
          response to drug
          glucose homeostasis
          response to ethanol
          glycerol-3-phosphate biosynthetic process
          glycerol kinase activity
          ATP binding
          histone binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P123949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19029 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111495 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82148 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84010 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109885 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39908 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 724 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173757 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120516 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170453 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123949 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123949 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P123949 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P148071 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60146 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23927 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104498 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4460 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39541 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10938240 - -
          Affymetrix Rat Gene 1.0 ST Array 10938242 - -
          Affymetrix Rat Gene 1.0 ST Array 10938227 - -
          Affymetrix Rat Gene 1.0 ST Array 10938236 - -
          Affymetrix Rat Gene 1.0 ST Array 10938231 - -
          Affymetrix Rat Gene 1.0 ST Array 10938232 - -
          Affymetrix Rat Gene 1.0 ST Array 10938229 - -
          Affymetrix Rat Gene 1.0 ST Array 10938221 - -
          Affymetrix Rat Gene 1.0 ST Array 10938230 - -
          Affymetrix Rat Gene 1.0 ST Array 10938226 - -
          Affymetrix Rat Gene 1.0 ST Array 10938225 - -
          Affymetrix Rat Gene 1.0 ST Array 10938238 - -
          Affymetrix Rat Gene 1.0 ST Array 10938233 - -
          Affymetrix Rat Gene 1.0 ST Array 10938235 - -
          Affymetrix Rat Gene 1.0 ST Array 10938228 - -
          Affymetrix Rat Gene 1.0 ST Array 10938239 - -
          Affymetrix Rat Gene 1.0 ST Array 10938234 - -
          Affymetrix Rat Gene 1.0 ST Array 10938220 - -
          Affymetrix Rat Gene 1.0 ST Array 10938224 - -
          Affymetrix Rat Gene 1.0 ST Array 10938241 - -
          Affymetrix Rat Gene 1.0 ST Array 10938223 - -
          Affymetrix Rat Gene 1.1 ST Array 10938219 - -
          Rat Genome 230 2.0 Array 1391661_at - -
          Rat Genome 230 2.0 Array 1387491_PM_at - -
          Rat Genome 230 2.0 Array 1387491_at - -
          Rat Genome 230 2.0 Array 1391661_PM_at - -
          Affymetrix HT Rat Focus Array 1387491_PM_at - -
          Affymetrix HT Rat Focus Array 1387491_at - -
          Affymetrix HT Rat Focus Array 1391661_PM_at - -
          Affymetrix HT Rat Focus Array 1391661_at - -
          Affymetrix Rat Clariom S Assay PSR0X00011506.rn.1 TC0X00001797.rn.1 RTA TC
          Rat Expression Set 230 1387491_at - -
          Rat Expression Set 230 1387491_PM_at - -
          Rat Expression Set 230 1391661_PM_at - -
          Rat Expression Set 230 1391661_at - -
          Affymetrix Rat Gene 1.0 ST Array 10938219 - -
          Rat Genome U34 Set D16102_at - -
          Rat Genome U34 Set rc_AA945076_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21089782 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline