Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577747_m1
          See other SERPIND1 GE Assays ›
          Gene Symbol:
          Serpind1
          Gene Name:
          serpin peptidase inhibitor, clade D (heparin cofactor), member 1
          Gene Aliases:
          rls2var1
          Chromosome Location:
          Chr.11: 87913814 - 87924880 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577747_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpind1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024382.1 NP_077358.1 4 - 5 1217 96
          XM_006248709.2 XP_006248771.1 3 - 4 1224 96
          GenBank mRNA BC091145.1 - 4 - 5 1197 7372906 96
          FQ210385.1 - 1 - 2 250 96
          FQ218607.1 - 3 - 4 1194 96
          X74549.1 - 4 - 5 1217 96
          X74550.1 - 3 - 4 1164 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100002013.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpind1

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          blood coagulation
          biological_process
          negative regulation of endopeptidase activity
          serine-type endopeptidase inhibitor activity
          heparin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P721883 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P721883 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P721883 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P721883 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P721883 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10755767 - -
          Affymetrix Rat Gene 1.0 ST Array 10755764 - -
          Affymetrix Rat Gene 1.0 ST Array 10755766 - -
          Affymetrix Rat Gene 1.0 ST Array 10755768 - -
          Affymetrix Rat Gene 1.0 ST Array 10755765 - -
          Affymetrix Rat Gene 1.0 ST Array 10755769 - -
          Affymetrix Rat Gene 1.1 ST Array 10755763 - -
          Rat Genome 230 2.0 Array 1398260_PM_a_at - -
          Rat Genome 230 2.0 Array 1398260_a_at - -
          Affymetrix HT Rat Focus Array 1398260_a_at - -
          Affymetrix HT Rat Focus Array 1398260_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1100013291.rn.1 TC1100002013.rn.1 RTA TC
          Rat Expression Set 230 1398260_a_at - -
          Rat Expression Set 230 1398260_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10755763 - -
          Rat Genome U34 Set X74549_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22065861 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352492 - -
          RatRef-12 Expression BeadChip ILMN_65813 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline