Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577775_m1
          See other HPGD GE Assays ›
          Gene Symbol:
          Hpgd
          Gene Name:
          hydroxyprostaglandin dehydrogenase 15 (NAD)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 37457134 - 37495758 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577775_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hpgd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024390.2 NP_077366.2 4 - 5 440 83
          GenBank mRNA BC062399.1 - 4 - 5 440 5599288 83
          U44750.1 - 4 - 5 456 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001566.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hpgd

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin metabolic process
          transforming growth factor beta receptor signaling pathway
          female pregnancy
          parturition
          ovulation
          negative regulation of cell cycle
          oxidation-reduction process
          thrombin receptor signaling pathway
          ductus arteriosus closure
          catalytic activity
          prostaglandin E receptor activity
          15-hydroxyprostaglandin dehydrogenase (NAD+) activity
          NAD binding
          NAD+ binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725524 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156951 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7373 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11511 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36718 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42877 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11278 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11278 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11278 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11278 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1500 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72422 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67964 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10791527 - -
          Affymetrix Rat Gene 1.0 ST Array 10791526 - -
          Affymetrix Rat Gene 1.0 ST Array 10791530 - -
          Affymetrix Rat Gene 1.0 ST Array 10791523 - -
          Affymetrix Rat Gene 1.0 ST Array 10791529 - -
          Affymetrix Rat Gene 1.0 ST Array 10791525 - -
          Affymetrix Rat Gene 1.0 ST Array 10791524 - -
          Affymetrix Rat Gene 1.0 ST Array 10791528 - -
          Affymetrix Rat Gene 1.1 ST Array 10791522 - -
          Rat Genome 230 2.0 Array 1376191_PM_at - -
          Rat Genome 230 2.0 Array 1376191_at - -
          Rat Genome 230 2.0 Array 1387631_at - -
          Rat Genome 230 2.0 Array 1387631_PM_at - -
          Affymetrix HT Rat Focus Array 1387631_PM_at - -
          Affymetrix HT Rat Focus Array 1376191_PM_at - -
          Affymetrix HT Rat Focus Array 1387631_at - -
          Affymetrix HT Rat Focus Array 1376191_at - -
          Affymetrix Rat Clariom S Assay PSR1600011881.rn.1 TC1600001566.rn.1 RTA TC
          Rat Expression Set 230 1376191_at - -
          Rat Expression Set 230 1387631_PM_at - -
          Rat Expression Set 230 1376191_PM_at - -
          Rat Expression Set 230 1387631_at - -
          Affymetrix Rat Gene 1.0 ST Array 10791522 - -
          Rat Genome U34 Set U44750_at - -
          Rat Genome U34 Set rc_AA848820_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21233763 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367170 - -
          RatRef-12 Expression BeadChip ILMN_53529 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline