Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577876_m1
          See other ACO2 GE Assays ›
          Gene Symbol:
          Aco2
          Gene Name:
          aconitase 2, mitochondrial
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 123102493 - 123145608 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577876_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aco2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024398.2 NP_077374.2 3 - 4 438 91
          GenBank mRNA AJ243266.1 - 3 - 4 439 91
          BC061999.1 - 3 - 4 438 5621226 91
          FQ216755.1 - 3 - 4 463 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001509.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aco2

          Panther Classification:

          Molecular Function -

          hydratase

          Gene Ontology Categories:

          Function(s) Process(es)

          tricarboxylic acid cycle
          citrate metabolic process
          isocitrate metabolic process
          aconitate hydratase activity
          iron ion binding
          protein binding
          3 iron, 4 sulfur cluster binding
          4 iron, 4 sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P158928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172340 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70219 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64242 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30078 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166233 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144589 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131153 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61456 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142225 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158928 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158928 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P158928 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P198349 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701612296 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73404 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70251 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 910 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6816 - -
          Agilent Rat cDNA Microarray (G4105A) 701612296 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10898018 - -
          Affymetrix Rat Gene 1.0 ST Array 10898009 - -
          Affymetrix Rat Gene 1.0 ST Array 10898010 - -
          Affymetrix Rat Gene 1.0 ST Array 10898007 - -
          Affymetrix Rat Gene 1.0 ST Array 10898019 - -
          Affymetrix Rat Gene 1.0 ST Array 10898006 - -
          Affymetrix Rat Gene 1.0 ST Array 10898016 - -
          Affymetrix Rat Gene 1.0 ST Array 10898005 - -
          Affymetrix Rat Gene 1.0 ST Array 10898008 - -
          Affymetrix Rat Gene 1.0 ST Array 10898004 - -
          Affymetrix Rat Gene 1.0 ST Array 10898021 - -
          Affymetrix Rat Gene 1.0 ST Array 10898017 - -
          Affymetrix Rat Gene 1.0 ST Array 10898011 - -
          Affymetrix Rat Gene 1.0 ST Array 10898020 - -
          Affymetrix Rat Gene 1.0 ST Array 10898014 - -
          Affymetrix Rat Gene 1.0 ST Array 10898013 - -
          Affymetrix Rat Gene 1.0 ST Array 10898012 - -
          Affymetrix Rat Gene 1.0 ST Array 10898015 - -
          Affymetrix Rat Gene 1.1 ST Array 10898003 - -
          Rat Genome 230 2.0 Array 1367589_PM_at - -
          Rat Genome 230 2.0 Array 1367589_at - -
          Affymetrix HT Rat Focus Array 1367589_PM_at - -
          Affymetrix HT Rat Focus Array 1367589_at - -
          Affymetrix Rat Clariom S Assay PSR0700011491.rn.1 TC0700001509.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700011493.rn.1 TC0700001509.rn.1 RTA TC
          Rat Expression Set 230 1367589_PM_at - -
          Rat Expression Set 230 1367589_at - -
          Affymetrix Rat Gene 1.0 ST Array 10898003 - -
          Rat Genome U34 Set rc_AI103956_at - -
          Rat Genome U34 Set rc_AI170777_at - -
          Rat Genome U34 Set rc_AI230919_at - -
          Rat Genome U34 Set rc_AI170777_g_at - -
          Rat Genome U34 Set rc_AI235320_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21496728 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352530 - -
          RatRef-12 Expression BeadChip ILMN_48841 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline