Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn00577929_m1
See other AXIN1 GE Assays ›
Gene Symbol:
Axin1
Gene Name:
axin 1
Gene Aliases:
Axin
Chromosome Location:
-
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn00577929_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Axin1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_024405.1
NP_077381.1
2409
56
GenBank mRNA
AF017756.1
-
2409
56
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
Recommended Assay
Rattus norvegicus
Rat Wnt Panel
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Axin1
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
in utero embryonic development
optic placode formation
regulation of protein phosphorylation
positive regulation of protein phosphorylation
nucleocytoplasmic transport
apoptotic process
activation of JUN kinase activity
determination of left/right symmetry
sensory perception of sound
adult walking behavior
cell death
dorsal/ventral axis specification
dorsal/ventral pattern formation
Wnt signaling pathway
hemoglobin metabolic process
forebrain anterior/posterior pattern specification
Wnt-activated signaling pathway involved in forebrain neuron fate commitment
protein catabolic process
negative regulation of Wnt signaling pathway
positive regulation of transforming growth factor beta receptor signaling pathway
olfactory placode formation
cytoplasmic microtubule organization
positive regulation of protein ubiquitination
activation of protein kinase activity
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
erythrocyte homeostasis
negative regulation of transcription elongation from RNA polymerase II promoter
regulation of catenin import into nucleus
post-anal tail morphogenesis
termination of G-protein coupled receptor signaling pathway
positive regulation of JUN kinase activity
positive regulation of GTPase activity
cellular protein complex assembly
negative regulation of fat cell differentiation
positive regulation of protein catabolic process
positive regulation of protein kinase activity
positive regulation of transcription, DNA-templated
positive regulation of JNK cascade
embryonic eye morphogenesis
axial mesoderm development
axial mesoderm formation
neurological system process
negative regulation of protein metabolic process
protein homooligomerization
positive regulation of ubiquitin-protein transferase activity
muscle cell development
canonical Wnt signaling pathway
embryonic skeletal joint morphogenesis
canonical Wnt signaling pathway involved in neural plate anterior/posterior pattern formation
cellular response to organic cyclic compound
genetic imprinting
negative regulation of canonical Wnt signaling pathway
Wnt signaling pathway involved in somitogenesis
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
p53 binding
signal transducer activity
GTPase activator activity
receptor binding
protein binding
beta-catenin binding
protein C-terminus binding
enzyme binding
protein kinase binding
protein domain specific binding
receptor signaling complex scaffold activity
ubiquitin protein ligase binding
protein complex scaffold
identical protein binding
protein homodimerization activity
protein self-association
SMAD binding
armadillo repeat domain binding
I-SMAD binding
R-SMAD binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline