Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578115_m1
          See other SLC16A3 GE Assays ›
          Gene Symbol:
          Slc16a3
          Gene Name:
          solute carrier family 16 (monocarboxylate transporter), member 3
          Gene Aliases:
          MCT4, Mct3
          Chromosome Location:
          Chr.10: 110138523 - 110148360 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578115_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc16a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030834.1 NP_110461.1 4 - 5 1229 86
          XM_006247931.2 XP_006247993.1 5 - 6 1344 86
          XM_006247932.2 XP_006247994.1 5 - 6 1636 86
          XM_006247933.2 XP_006247995.1 4 - 5 1251 86
          XM_006247934.2 XP_006247996.1 5 - 6 1310 86
          XM_008768513.1 XP_008766735.1 5 - 6 4068 86
          XM_008768514.1 XP_008766736.1 4 - 5 3975 86
          GenBank mRNA BC168146.1 - 4 - 5 1188 9089640 86
          U87627.1 - 4 - 5 1229 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002097.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc16a3

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          plasma membrane lactate transport
          lactate transmembrane transporter activity
          symporter activity
          secondary active monocarboxylate transmembrane transporter activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373041 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122986 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12451 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12451 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12451 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12451 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10740212 - -
          Affymetrix Rat Gene 1.0 ST Array 10740210 - -
          Affymetrix Rat Gene 1.0 ST Array 10740213 - -
          Affymetrix Rat Gene 1.0 ST Array 10749743 - -
          Affymetrix Rat Gene 1.0 ST Array 10740211 - -
          Affymetrix Rat Gene 1.1 ST Array 10749742 - -
          Affymetrix Rat Gene 1.1 ST Array 10740209 - -
          Rat Genome 230 2.0 Array 1368965_at - -
          Rat Genome 230 2.0 Array 1368965_PM_at - -
          Affymetrix HT Rat Focus Array 1368965_PM_at - -
          Affymetrix HT Rat Focus Array 1368965_at - -
          Affymetrix Rat Clariom S Assay PSR1000019013.rn.1 TC1000002097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000019017.rn.1 TC1000002097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000019016.rn.1 TC1000002097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000019014.rn.1 TC1000002097.rn.1 RTA TC
          Rat Expression Set 230 1368965_at - -
          Rat Expression Set 230 1368965_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10740209 - -
          Rat Genome U34 Set U87627_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21796082 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350798 - -
          RatRef-12 Expression BeadChip ILMN_64676 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline