Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578116_m1
          See other SERP1 GE Assays ›
          Gene Symbol:
          Serp1
          Gene Name:
          stress-associated endoplasmic reticulum protein 1
          Gene Aliases:
          RAMP4
          Chromosome Location:
          Chr.2: 148718279 - 148722263 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578116_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030835.2 NP_110462.1 2 - 3 522 81
          GenBank mRNA AB018546.1 - 2 - 3 407 81
          AF100470.1 - 2 - 3 176 81
          AJ238236.1 - 2 - 3 182 81
          BC061854.1 - 2 - 3 522 5621665 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003747.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serp1

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s)

          skeletal system development
          glucose metabolic process
          protein glycosylation
          post-embryonic development
          multicellular organism aging
          protein transport
          endoplasmic reticulum unfolded protein response
          positive regulation of insulin secretion
          positive regulation of translation
          positive regulation of organ growth
          muscle organ morphogenesis
          positive regulation of growth hormone secretion

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P193990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470938 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P193990 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P193990 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P193990 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P193990 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700323002 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4339 - -
          Agilent Rat cDNA Microarray (G4105A) 700323002 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823314 - -
          Affymetrix Rat Gene 1.0 ST Array 10823310 - -
          Affymetrix Rat Gene 1.0 ST Array 10823312 - -
          Affymetrix Rat Gene 1.0 ST Array 10823311 - -
          Affymetrix Rat Gene 1.0 ST Array 10823313 - -
          Affymetrix Rat Gene 1.0 ST Array 10823315 - -
          Affymetrix Rat Gene 1.1 ST Array 10823309 - -
          Rat Genome 230 2.0 Array 1386900_PM_at - -
          Rat Genome 230 2.0 Array 1386900_at - -
          Rat Genome 230 2.0 Array 1367686_at - -
          Rat Genome 230 2.0 Array 1367686_PM_at - -
          Affymetrix HT Rat Focus Array 1386900_at - -
          Affymetrix HT Rat Focus Array 1386900_PM_at - -
          Affymetrix HT Rat Focus Array 1367686_PM_at - -
          Affymetrix HT Rat Focus Array 1367686_at - -
          Affymetrix Rat Clariom S Assay PSR0200025098.rn.1 TC0200003747.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200025099.rn.1 TC0200003747.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200025097.rn.1 TC0200003747.rn.1 RTA TC
          Rat Expression Set 230 1367686_PM_at - -
          Rat Expression Set 230 1386900_at - -
          Rat Expression Set 230 1367686_at - -
          Rat Expression Set 230 1386900_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10823309 - -
          Rat Genome U34 Set AF100470_at - -
          Rat Genome U34 Set AF100470_g_at - -
          Rat Genome U34 Set rc_AI231302_at - -
          Rat Genome U34 Set rc_AI103695_r_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22013907 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354484 - -
          RatRef-12 Expression BeadChip ILMN_50573 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline