Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578125_m1
          See other ERAP1 GE Assays ›
          Gene Symbol:
          Erap1
          Gene Name:
          endoplasmic reticulum aminopeptidase 1
          Gene Aliases:
          Appils, Arts1
          Chromosome Location:
          Chr.2: 1410888 - 1449734 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578125_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Erap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030836.1 NP_110463.1 2 - 3 693 60
          XM_006231700.1 XP_006231762.1 3 - 4 1427 60
          XM_006231701.2 XP_006231763.1 3 - 4 746 60
          XM_006231704.2 XP_006231766.1 3 - 4 746 60
          XM_008760600.1 XP_008758822.1 2 - 3 2839 60
          XM_008760601.1 XP_008758823.1 2 - 3 2840 60
          XM_008760602.1 XP_008758824.1 2 - 3 2838 60
          GenBank mRNA AF148323.1 - 2 - 3 690 60
          AF148324.1 - 2 - 3 693 60
          BC098664.1 - 3 - 4 700 7192921 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000006.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Erap1

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          membrane protein ectodomain proteolysis
          response to bacterium
          antigen processing and presentation
          positive regulation of angiogenesis
          aminopeptidase activity
          interleukin-6 receptor binding
          tumor necrosis factor receptor binding
          peptidase activity
          metalloexopeptidase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136805 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119070 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6581 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153195 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141337 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88236 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15303 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15303 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15303 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15303 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700307443 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65916 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23036 - -
          Agilent Rat cDNA Microarray (G4105A) 700307443 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10812170 - -
          Affymetrix Rat Gene 1.0 ST Array 10812181 - -
          Affymetrix Rat Gene 1.0 ST Array 10812164 - -
          Affymetrix Rat Gene 1.0 ST Array 10812180 - -
          Affymetrix Rat Gene 1.0 ST Array 10812175 - -
          Affymetrix Rat Gene 1.0 ST Array 10812178 - -
          Affymetrix Rat Gene 1.0 ST Array 10812177 - -
          Affymetrix Rat Gene 1.0 ST Array 10812172 - -
          Affymetrix Rat Gene 1.0 ST Array 10812176 - -
          Affymetrix Rat Gene 1.0 ST Array 10812165 - -
          Affymetrix Rat Gene 1.0 ST Array 10812167 - -
          Affymetrix Rat Gene 1.0 ST Array 10812173 - -
          Affymetrix Rat Gene 1.0 ST Array 10812169 - -
          Affymetrix Rat Gene 1.0 ST Array 10812171 - -
          Affymetrix Rat Gene 1.0 ST Array 10812179 - -
          Affymetrix Rat Gene 1.0 ST Array 10812166 - -
          Affymetrix Rat Gene 1.0 ST Array 10812174 - -
          Affymetrix Rat Gene 1.0 ST Array 10812163 - -
          Affymetrix Rat Gene 1.0 ST Array 10812168 - -
          Affymetrix Rat Gene 1.1 ST Array 10812162 - -
          Rat Genome 230 2.0 Array 1368356_PM_a_at - -
          Rat Genome 230 2.0 Array 1399161_PM_a_at - -
          Rat Genome 230 2.0 Array 1387149_at - -
          Rat Genome 230 2.0 Array 1387149_PM_at - -
          Rat Genome 230 2.0 Array 1368356_a_at - -
          Rat Genome 230 2.0 Array 1399161_a_at - -
          Affymetrix HT Rat Focus Array 1368356_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387149_PM_at - -
          Affymetrix HT Rat Focus Array 1399161_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387149_at - -
          Affymetrix HT Rat Focus Array 1399161_a_at - -
          Affymetrix HT Rat Focus Array 1368356_a_at - -
          Affymetrix Rat Clariom S Assay PSR0200000020.rn.1 TC0200000006.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000022.rn.1 TC0200000006.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000021.rn.1 TC0200000006.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000019.rn.1 TC0200000006.rn.1 RTA TC
          Rat Expression Set 230 1399161_PM_a_at - -
          Rat Expression Set 230 1387149_at - -
          Rat Expression Set 230 1399161_a_at - -
          Rat Expression Set 230 1368356_a_at - -
          Rat Expression Set 230 1387149_PM_at - -
          Rat Expression Set 230 1368356_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10812162 - -
          Rat Genome U34 Set rc_AI044567_g_at - -
          Rat Genome U34 Set rc_AI044567_at - -
          Rat Genome U34 Set rc_AI171503_at - -
          Rat Genome U34 Set rc_AI170071_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21161241 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368616 - -
          RatRef-12 Expression BeadChip ILMN_52339 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline