Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578208_m1
          See other STXBP5 GE Assays ›
          Gene Symbol:
          Stxbp5
          Gene Name:
          syntaxin binding protein 5 (tomosyn)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 3858917 - 4011688 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578208_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stxbp5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030843.2 NP_110470.1 8 - 9 1194 68
          NM_178345.1 NP_848035.1 8 - 9 844 68
          NM_178346.1 NP_848036.1 8 - 9 844 68
          XM_006227623.2 XP_006227685.1 8 - 9 1371 68
          XM_006227624.2 XP_006227686.1 8 - 9 1371 68
          XM_008758628.1 XP_008756850.1 8 - 9 1371 68
          XM_008758629.1 XP_008756851.1 8 - 9 1371 68
          GenBank mRNA AF118889.1 - 8 - 9 844 68
          AF118890.1 - 8 - 9 844 68
          U92072.1 - 8 - 9 1194 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003990.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stxbp5

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          regulation of gene expression
          protein transport
          regulation of exocytosis
          regulation of blood coagulation
          positive regulation of exocytosis
          syntaxin-1 binding
          syntaxin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P163189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640754 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157373 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48619 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149252 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119635 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89980 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132225 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26391 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118431 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74674 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32712 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50795 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158498 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28360 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19277 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54904 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115560 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144821 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P163189 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P163189 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P163189 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P163189 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700520251 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104390 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95686 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98258 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56833 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94127 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81751 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29054 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103663 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54372 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48992 - -
          Agilent Rat cDNA Microarray (G4105A) 700520251 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10716713 - -
          Affymetrix Rat Gene 1.0 ST Array 10716719 - -
          Affymetrix Rat Gene 1.0 ST Array 10716714 - -
          Affymetrix Rat Gene 1.0 ST Array 10716734 - -
          Affymetrix Rat Gene 1.0 ST Array 10716725 - -
          Affymetrix Rat Gene 1.0 ST Array 10716738 - -
          Affymetrix Rat Gene 1.0 ST Array 10716733 - -
          Affymetrix Rat Gene 1.0 ST Array 10716731 - -
          Affymetrix Rat Gene 1.0 ST Array 10716722 - -
          Affymetrix Rat Gene 1.0 ST Array 10716739 - -
          Affymetrix Rat Gene 1.0 ST Array 10716724 - -
          Affymetrix Rat Gene 1.0 ST Array 10716715 - -
          Affymetrix Rat Gene 1.0 ST Array 10716726 - -
          Affymetrix Rat Gene 1.0 ST Array 10716736 - -
          Affymetrix Rat Gene 1.0 ST Array 10716740 - -
          Affymetrix Rat Gene 1.0 ST Array 10716727 - -
          Affymetrix Rat Gene 1.0 ST Array 10716728 - -
          Affymetrix Rat Gene 1.0 ST Array 10716723 - -
          Affymetrix Rat Gene 1.0 ST Array 10716721 - -
          Affymetrix Rat Gene 1.0 ST Array 10716716 - -
          Affymetrix Rat Gene 1.0 ST Array 10716720 - -
          Affymetrix Rat Gene 1.0 ST Array 10716729 - -
          Affymetrix Rat Gene 1.0 ST Array 10716735 - -
          Affymetrix Rat Gene 1.0 ST Array 10716741 - -
          Affymetrix Rat Gene 1.0 ST Array 10716730 - -
          Affymetrix Rat Gene 1.0 ST Array 10716732 - -
          Affymetrix Rat Gene 1.0 ST Array 10716717 - -
          Affymetrix Rat Gene 1.0 ST Array 10716718 - -
          Affymetrix Rat Gene 1.0 ST Array 10716737 - -
          Affymetrix Rat Gene 1.1 ST Array 10716712 - -
          Rat Genome 230 2.0 Array 1387415_PM_a_at - -
          Rat Genome 230 2.0 Array 1393096_PM_at - -
          Rat Genome 230 2.0 Array 1387415_a_at - -
          Rat Genome 230 2.0 Array 1393096_at - -
          Affymetrix HT Rat Focus Array 1387415_a_at - -
          Affymetrix HT Rat Focus Array 1387415_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100032424.rn.1 TC0100003990.rn.1 RTA TC
          Rat Expression Set 230 1387415_a_at - -
          Rat Expression Set 230 1387415_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10716712 - -
          Rat Genome U34 Set U92072_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20929555 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375508 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376692 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376134 - -
          RatRef-12 Expression BeadChip ILMN_60777 - -
          RatRef-12 Expression BeadChip ILMN_62118 - -
          RatRef-12 Expression BeadChip ILMN_54781 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline