Search
Search
| Gene Symbol: Emp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030847.1 | NP_110474.1 | 4 - 5 | 523 | 118 | |
| XM_006229158.2 | XP_006229220.1 | 4 - 5 | 587 | 118 | ||
| XM_008759418.1 | XP_008757640.1 | 4 - 5 | 465 | 118 | ||
| GenBank mRNA | BC088131.1 | - | 4 - 5 | 519 | 7365466 | 118 |
| FQ233038.1 | - | 4 - 5 | 510 | 118 | ||
| Y10889.2 | - | 4 - 5 | 523 | 118 | ||
Set Membership: |
|