Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578255_m1
          See other BHMT GE Assays ›
          Gene Symbol:
          Bhmt
          Gene Name:
          betaine-homocysteine S-methyltransferase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 23236573 - 23256158 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578255_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bhmt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030850.1 NP_110477.1 2 - 3 233 116
          GenBank mRNA AF038870.1 - 2 - 3 233 116
          BC078811.1 - 2 - 3 220 7130736 116
          U96133.1 - 2 - 3 222 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002619.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bhmt

          Panther Classification:

          Molecular Function -

          methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein methylation
          amino-acid betaine metabolic process
          amino-acid betaine catabolic process
          methionine biosynthetic process
          response to organonitrogen compound
          S-methylmethionine cycle
          S-adenosylmethionine metabolic process
          L-methionine salvage
          methyltransferase activity
          zinc ion binding
          S-adenosylmethionine-homocysteine S-methyltransferase activity
          protein complex binding
          betaine-homocysteine S-methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P535284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98203 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71573 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P535284 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P535284 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P535284 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P360510 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P535284 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700607442 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104724 - -
          Agilent Rat cDNA Microarray (G4105A) 700607442 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10820495 - -
          Affymetrix Rat Gene 1.0 ST Array 10820498 - -
          Affymetrix Rat Gene 1.0 ST Array 10820497 - -
          Affymetrix Rat Gene 1.0 ST Array 10820499 - -
          Affymetrix Rat Gene 1.0 ST Array 10820501 - -
          Affymetrix Rat Gene 1.0 ST Array 10820500 - -
          Affymetrix Rat Gene 1.0 ST Array 10820496 - -
          Affymetrix Rat Gene 1.1 ST Array 10820494 - -
          Rat Genome 230 2.0 Array 1387665_at - -
          Rat Genome 230 2.0 Array 1387665_PM_at - -
          Affymetrix HT Rat Focus Array 1387665_at - -
          Affymetrix HT Rat Focus Array 1387665_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200018363.rn.1 TC0200002619.rn.1 RTA TC
          Rat Expression Set 230 1387665_at - -
          Rat Expression Set 230 1387665_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10820494 - -
          Rat Genome U34 Set AF038870_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21165857 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039973 - -
          RatRef-12 Expression BeadChip ILMN_50334 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline