Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578277_m1
          See other LECT1 GE Assays ›
          Gene Symbol:
          Lect1
          Gene Name:
          leukocyte cell derived chemotaxin 1
          Gene Aliases:
          Chm-1
          Chromosome Location:
          Chr.15: 61923291 - 61947764 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578277_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lect1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030854.1 NP_110481.1 3 - 4 488 72
          XM_006252346.1 XP_006252408.1 3 - 4 483 72
          GenBank mRNA AF051425.1 - 3 - 4 488 72
          BC127493.1 - 3 - 4 429 8369609 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500002151.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lect1

          Gene Ontology Categories:

          Function(s)

          endothelial cell morphogenesis
          negative regulation of endothelial cell proliferation
          negative regulation of angiogenesis
          cell differentiation
          negative regulation of vascular endothelial growth factor receptor signaling pathway
          cartilage development

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P631818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424158 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103200 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85197 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P631818 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P631818 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P631818 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P631818 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79547 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39493 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10785453 - -
          Affymetrix Rat Gene 1.0 ST Array 10785458 - -
          Affymetrix Rat Gene 1.0 ST Array 10785456 - -
          Affymetrix Rat Gene 1.0 ST Array 10785460 - -
          Affymetrix Rat Gene 1.0 ST Array 10785459 - -
          Affymetrix Rat Gene 1.0 ST Array 10785457 - -
          Affymetrix Rat Gene 1.0 ST Array 10785454 - -
          Affymetrix Rat Gene 1.0 ST Array 10785455 - -
          Affymetrix Rat Gene 1.1 ST Array 10785452 - -
          Rat Genome 230 2.0 Array 1387164_PM_at - -
          Rat Genome 230 2.0 Array 1387164_at - -
          Affymetrix HT Rat Focus Array 1387164_PM_at - -
          Affymetrix HT Rat Focus Array 1387164_at - -
          Affymetrix Rat Clariom S Assay PSR1500014799.rn.1 TC1500002151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500014801.rn.1 TC1500002151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500014794.rn.1 TC1500002151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500014798.rn.1 TC1500002151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500014802.rn.1 TC1500002151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500014793.rn.1 TC1500002151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500014800.rn.1 TC1500002151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500014796.rn.1 TC1500002151.rn.1 RTA TC
          Rat Expression Set 230 1387164_at - -
          Rat Expression Set 230 1387164_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10785452 - -
          Rat Genome U34 Set AF051425_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21643715 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372067 - -
          RatRef-12 Expression BeadChip ILMN_48761 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline