Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578382_m1
          See other MYOC GE Assays ›
          Gene Symbol:
          Myoc
          Gene Name:
          myocilin, trabecular meshwork inducible glucocorticoid response
          Gene Aliases:
          Tigr
          Chromosome Location:
          Chr.13: 80517531 - 80527928 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578382_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myoc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030865.1 NP_110492.1 1 - 2 684 70
          GenBank mRNA AB019393.1 - 1 - 2 680 70
          AF093567.1 - 1 - 2 684 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000629.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myoc

          Panther Classification:

          Molecular Function -

          structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          negative regulation of cell-matrix adhesion
          positive regulation of phosphatidylinositol 3-kinase signaling
          skeletal muscle hypertrophy
          nerve development
          myelination in peripheral nervous system
          positive regulation of cell migration
          neuron projection development
          negative regulation of Rho protein signal transduction
          non-canonical Wnt signaling pathway via JNK cascade
          ERBB2-ERBB3 signaling pathway
          regulation of MAPK cascade
          clustering of voltage-gated sodium channels
          positive regulation of stress fiber assembly
          negative regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          positive regulation of protein kinase B signaling
          positive regulation of mitochondrial depolarization
          bone development
          positive regulation of substrate adhesion-dependent cell spreading
          cellular response to nerve growth factor stimulus
          fibronectin binding
          molecular_function
          frizzled binding
          receptor tyrosine kinase binding
          myosin light chain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P547246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581879 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P547246 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P547246 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P547246 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P547246 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74250 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10765126 - -
          Affymetrix Rat Gene 1.0 ST Array 10765125 - -
          Affymetrix Rat Gene 1.0 ST Array 10765127 - -
          Affymetrix Rat Gene 1.1 ST Array 10765124 - -
          Rat Genome 230 2.0 Array 1387313_PM_at - -
          Rat Genome 230 2.0 Array 1387313_at - -
          Affymetrix HT Rat Focus Array 1387313_at - -
          Affymetrix HT Rat Focus Array 1387313_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300004379.rn.1 TC1300000629.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004380.rn.1 TC1300000629.rn.1 RTA TC
          Rat Expression Set 230 1387313_PM_at - -
          Rat Expression Set 230 1387313_at - -
          Affymetrix Rat Gene 1.0 ST Array 10765124 - -
          Rat Genome U34 Set AB019393_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22354353 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376768 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline