Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578384_m1
          See other NFKBIB GE Assays ›
          Gene Symbol:
          Nfkbib
          Gene Name:
          nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 86941073 - 86948845 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578384_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nfkbib
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030867.2 NP_110494.2 1 - 2 438 59
          GenBank mRNA AF246634.1 - 1 - 2 261 59
          BC085729.1 - 1 - 2 438 7114624 59
          FQ228042.1 - 1 - 2 235 59
          FQ235103.1 - 1 - 2 383 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005090.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nfkbib

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          cytoplasmic sequestering of NF-kappaB
          cellular response to lipopolysaccharide
          NF-kappaB binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P186860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616921 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141874 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P186860 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P186860 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P186860 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P186860 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700504017 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88553 - -
          Agilent Rat cDNA Microarray (G4105A) 700504017 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10720265 - -
          Affymetrix Rat Gene 1.0 ST Array 10720267 - -
          Affymetrix Rat Gene 1.0 ST Array 10720271 - -
          Affymetrix Rat Gene 1.0 ST Array 10720270 - -
          Affymetrix Rat Gene 1.0 ST Array 10720268 - -
          Affymetrix Rat Gene 1.0 ST Array 10720266 - -
          Affymetrix Rat Gene 1.0 ST Array 10720269 - -
          Affymetrix Rat Gene 1.0 ST Array 10720263 - -
          Affymetrix Rat Gene 1.1 ST Array 10720262 - -
          Rat Genome 230 2.0 Array 1367943_PM_at - -
          Rat Genome 230 2.0 Array 1367943_at - -
          Affymetrix HT Rat Focus Array 1367943_PM_at - -
          Affymetrix HT Rat Focus Array 1367943_at - -
          Affymetrix Rat Clariom S Assay PSR0100040513.rn.1 TC0100005090.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040517.rn.1 TC0100005090.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040514.rn.1 TC0100005090.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040518.rn.1 TC0100005090.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040515.rn.1 TC0100005090.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040516.rn.1 TC0100005090.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040512.rn.1 TC0100005090.rn.1 RTA TC
          Rat Expression Set 230 1367943_at - -
          Rat Expression Set 230 1367943_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10720262 - -
          Rat Genome U34 Set rc_AA925318_g_at - -
          Rat Genome U34 Set rc_AA925318_at - -
          Rat Genome U34 Set rc_AI176319_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21233750 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350438 - -
          RatRef-12 Expression BeadChip ILMN_69984 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline